Skip to Content
Merck
All Photos(1)

Key Documents

SMB00705

Sigma-Aldrich

Sesamin

≥98% (HPLC)

Synonym(s):

Fagarol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O6
CAS Number:
Molecular Weight:
354.35
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: 10 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C1Oc2ccc(cc2O1)[C@H]3OC[C@H]4[C@@H]3CO[C@@H]4c5ccc6OCOc6c5

InChI

1S/C20H18O6/c1-3-15-17(25-9-23-15)5-11(1)19-13-7-22-20(14(13)8-21-19)12-2-4-16-18(6-12)26-10-24-16/h1-6,13-14,19-20H,7-10H2/t13-,14-,19+,20+/m0/s1

InChI key

PEYUIKBAABKQKQ-AFHBHXEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sesamin is the most abundant lignan isolated from Sesamum indicum or sesame seeds and sesame oil.

Application

Sesamin has been used as a reference standard to study the activity of Cytochrome P450 enzyme sesamin synthase from Sesamum indicum L. in the conversion of pinoresinol to sesamin.

Biochem/physiol Actions

Non-competitive Δ5-desaturase inhibitor.
Sesamin presents multiple health benefits including hypocholesterolemic, anti-hypertensive, anti-carcinogenic, anti-aging, immunoregulatory, hypoglycemic, anti-thrombotic, hepatoprotective, and antioxidant activities. It has also been demonstrated to work synergistically with vitamin E against lipid peroxidation. Sesamin was found to be metabolized into several catechol forms in the liver. These cathecols have a potent ability to scavenge superoxide radicals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Dalibalta et al.
Saudi pharmaceutical journal : SPJ : the official publication of the Saudi Pharmaceutical Society, 28(10), 1276-1289 (2020-11-03)
Sesamin, a major lignin isolated from sesame (Sesamum indicum) seeds and sesame oil, is known to possess antioxidant and anti-inflammatory properties. Several studies have revealed that oxidative stress and inflammation play a major role in a variety of cardiovascular diseases
Mitsuo Namiki
Critical reviews in food science and nutrition, 47(7), 651-673 (2007-10-19)
The results of recent food chemical and nutraceutical studies on the traditionally well-known health value of sesame are described, including some aspects of the utilization of sesame. The highly antioxidative activity of sesame oil was clarified and found to involve
Colonoscope constriction and laser safety interlock problems investigated.
Today's OR nurse, 14(3), 48-48 (1992-03-01)
W C Johnson et al.
Life sciences, 51(15), 1229-1236 (1992-01-01)
A two step fractionation of conditioned media made from the darkly pigmented dorsal skin of the channel catfish, Ictalurus punctatus, has produced fractions that contain a melanization stimulating factor (MSF). Isolated neural tubes of Xenopus laevis embryos exposed to conditioned
Yun-Ping Lim et al.
Evidence-based complementary and alternative medicine : eCAM, 2012, 242810-242810 (2012-05-31)
Inconsistent expression and regulation of drug-metabolizing enzymes (DMEs) are common causes of adverse drug effects in some drugs with a narrow therapeutic index (TI). An important cytochrome, cytochrome P450 3A4 (CYP3A4), is predominantly regulated by a nuclear receptor, pregnane X

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service