Skip to Content
Merck
All Photos(1)

Key Documents

SMB00282

Sigma-Aldrich

Xylobiose

≥90% (HPLC)

Synonym(s):

Xylobiose, (2R,3R,4R)-2,3,5-Trihydroxy-4-[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxypentanal, 4-O-β-D-Xylopyranosyl-D-xylose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O9
CAS Number:
Molecular Weight:
282.24
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white to off-white

solubility

water: 10 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OCC(OC1OCC(O)C(O)C1O)C(O)C(O)C=O

InChI

1S/C10H18O9/c11-1-4(13)7(15)6(2-12)19-10-9(17)8(16)5(14)3-18-10/h1,4-10,12-17H,2-3H2

InChI key

SQNRKWHRVIAKLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A disaccharide

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cláudia Amorim et al.
Food chemistry, 270, 86-94 (2018-09-04)
Xylooligosaccharides (XOS) are prebiotic nutraceuticals that can be sourced from lignocellulosic biomass, such as agro-residues. This study reports for the first time an optimization study of XOS production from agro-residues by direct fermentation using two Trichoderma species. A total of
Jacob Krüger Jensen et al.
The New phytologist, 218(3), 1049-1060 (2018-02-21)
Insights into the evolution of plant cell walls have important implications for comprehending these diverse and abundant biological structures. In order to understand the evolving structure-function relationships of the plant cell wall, it is imperative to trace the origin of
Hao Shi et al.
Biotechnology for biofuels, 6(1), 26-26 (2013-02-20)
Xylanase is an important component of hemicellulase enzyme system. Since it plays an important role in the hydrolysis of hemicellulose into xylooligosaccharides (XOs), high thermostable xylanase has been the focus of much recent attention as powerful enzyme as well as
Bing Guo et al.
Marine drugs, 11(4), 1173-1187 (2013-04-10)
Marine xylanases are rather less studied compared to terrestrial xylanases. In this study, a new xylanase gene, xynB, was cloned from the marine bacterium, Glaciecola mesophila KMM241, and expressed in Escherichia coli. xynB encodes a multi-domain xylanase XynB of glycoside
Yaping Gao et al.
Plant physiology, 173(1), 470-481 (2016-11-20)
Acetylation is a ubiquitous modification on cell wall polymers, which play a structural role in plant growth and stress defenses. However, the mechanisms for how crop plants accomplish cell wall polymer O-acetylation are largely unknown. Here, we report on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service