Skip to Content
Merck

C8286

Cellotetraose

≥85% (HPLC)

Synonym(s):

(β-D-Glc-[1→4])3-D-Glc, D-(+)-Cellotetraose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H42O21
CAS Number:
Molecular Weight:
666.58
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
78869
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

assay

≥85% (HPLC)

form

powder

color

white

solubility

water: 25 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)C(O)O[C@@H]4CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C24H42O21/c25-1-5-9(29)10(30)15(35)22(40-5)44-19-7(3-27)42-24(17(37)12(19)32)45-20-8(4-28)41-23(16(36)13(20)33)43-18-6(2-26)39-21(38)14(34)11(18)31/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10+,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21?,22+,23+,24+/m1/s1

InChI key

LUEWUZLMQUOBSB-YQGOCCRESA-N

Application

Cellotetraose is a 4-glucose polymer cellulodextrin derived from cellulose degradation. Cellotetraose and other cellulodextrins, such as cellulobiose, cellulotriose and cellopentaose are used by cellulosic bacteria as sources of energy. Cellotetraose is used to help identify, differentiate and characterize oligosaccharide metabolizing enzymes.

Biochem/physiol Actions

Cellotetraose is a substrate for many cellulases and for 1,4-β-D-glucan glucohydrolases. It is also the end product of some cellulases, such as Cel9R from Clostridium thermocellum.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library