Skip to Content
Merck
All Photos(1)

Key Documents

SMB00082

Sigma-Aldrich

Albiflorin

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28O11
CAS Number:
Molecular Weight:
480.46
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C[C@]12C[C@H]3[C@H](O)C[C@@]1(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@]3(COC(=O)c5ccccc5)C(=O)O2

InChI

1S/C23H28O11/c1-21-7-12-13(25)8-23(21,33-19-17(28)16(27)15(26)14(9-24)32-19)22(12,20(30)34-21)10-31-18(29)11-5-3-2-4-6-11/h2-6,12-17,19,24-28H,7-10H2,1H3/t12-,13+,14+,15+,16-,17+,19-,21-,22-,23-/m0/s1

InChI key

JCLNQDVXOFIJKI-ICECTASOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Biochem/physiol Actions

Albiflorin, along with paeoniflorin, are the main components of Shaoyao-Gancao, a traditional Chinese medicine used for pain relief.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hee-Jung Sim et al.
Journal of chromatography. A, 1217(32), 5302-5305 (2010-07-06)
We developed a method to simultaneously determine paeoniflorin and albiflorin levels using high-performance anion-exchange liquid chromatography with pulsed amperometric detection (HPAEC-PAD). The main principle of our method includes solid-phase extraction step using Amberlite XAD-2 sorbent to remove sugars and to
Mi-Yeon Jung et al.
Biomolecules, 9(7) (2019-07-13)
Gami-soyosan is a medicinal herbal formulation prescribed for the treatment of menopausal symptoms, including hot flashes and osteoporosis. Gami-soyosan is also used to treat similar symptoms experienced by patients with breast cancer. The incidence of breast cancer in women receiving
Min-Hwan Jeon et al.
Journal of chromatography. A, 1216(21), 4568-4573 (2009-04-07)
We have developed a reversed-phase high-performance liquid chromatography-pulsed amperometric detection (RP-HPLC-PAD) method for the detection of albiflorin and paeoniflorin in Paeoniae Radix and Wu-ji-san. Albiflorin and paeoniflorin were completely separated using 10% acetonitrile in 5mM sodium phosphate buffer (pH 3.0)
Chao Feng et al.
Journal of ethnopharmacology, 130(2), 407-413 (2010-06-29)
To establish a HPLC-MS method and investigate the pharmacokinetic properties of paeoniflorin, albiflorin and oxypaeoniflorin and the pharmacokinetics difference of Radix Paeoniae Rubra and Radix Paeoniae Alba. The extracts of Radix Paeoniae Rubra and Radix Paeoniae Alba were separately administrated
Rui Li et al.
Journal of mass spectrometry : JMS, 42(3), 335-345 (2007-01-03)
In order to analyze paeoniflorin, albiflorin and their derivatives (PADs) in Paeonia Lactiflora rapidly and effectively, (+)ESI-MS(n) experiments were conducted, from which two diagnostic fragment patterns were acquired. Meanwhile, the dehydration ability of aglycones of PADs was obtained by calculating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service