Skip to Content
Merck
All Photos(1)

Key Documents

SML0054

Sigma-Aldrich

Schizandrin

≥98% (HPLC)

Synonym(s):

(+)-Schizandrin, Schisandrin, Schisandrine, Wuweizi alcohol A, Wuweizichun A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O7
CAS Number:
Molecular Weight:
432.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 75 to 95°, c = 1 in methanol

color

white to beige

solubility

DMSO: ≥13 mg/mL

storage temp.

room temp

SMILES string

COc1cc2C[C@H](C)[C@@](C)(O)Cc3cc(OC)c(OC)c(OC)c3-c2c(OC)c1OC

InChI

1S/C24H32O7/c1-13-9-14-10-16(26-3)20(28-5)22(30-7)18(14)19-15(12-24(13,2)25)11-17(27-4)21(29-6)23(19)31-8/h10-11,13,25H,9,12H2,1-8H3/t13-,24-/m0/s1

InChI key

YEFOAORQXAOVJQ-RZFZLAGVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Schizandrin may be used in oxidative stress-related cell signaling studies.

Biochem/physiol Actions

Antioxidant; Anti-inflammatory, reduces oxidative stress.
Schizandrin is a natural product with several biological properties involved with antioxidant and anti-inflammatory activities. It reduces the formation of ROS, inhibits the mitochondrial pathway of the apoptotic process and oxidative stress. Schizandrin has been reported to have hepatoprotective, antitumor, antiviral, and antiamnesic effects, and has neuroprotective activity against glutamate induced neurotoxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elina Hakala et al.
The Journal of antibiotics, 68(10), 609-614 (2015-05-07)
Lignans from Schisandra chinensis berries show various pharmacological activities, of which their antioxidative and cytoprotective properties are among the most studied ones. Here, the first report on antibacterial properties of six dibenzocyclooctadiene lignans found in Schisandra spp. is presented. The
Chun-hui Ma et al.
Analytica chimica acta, 689(1), 110-116 (2011-02-23)
The ionic liquid-based ultrasonic-assisted extraction (ILUAE) has been successfully applied in extracting four biphenyl cyclooctene lignans from the fruit of Schisandra chinensis Baill. Seventeen different types of ionic liquids with different cations and anions have been investigated. 0.8 M 1-lauryl-3-methylimidazolium
Jiaming Yang et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(11), 998-1005 (2011-04-26)
This study examined the effect of schisandrin, one of the major lignans isolated from Schisandra chinensis, on spontaneous contraction in rat colon and its possible mechanisms. Schisandrin produced a concentration-dependent inhibition (EC₅₀=1.66 μM) on the colonic spontaneous contraction. The relaxant
Di Hu et al.
Oxidative medicine and cellular longevity, 2012, 721721-721721 (2012-07-26)
In the present study, we examined the effect of schisandrin (SCH) of Schisandra chinensis on the amyloid-beta(1-42)- (Aβ(1-42)-) induced memory impairment in mice and elucidated the possible antioxidative mechanism. Mice were intracerebroventricular (i.c.v.) injected with the aggregated Aβ(1-42) and then
Ju-Xian Song et al.
Phytotherapy research : PTR, 25(3), 435-443 (2010-08-27)
Aggregated beta-amyloid (Aβ) and elevated plasma levels of homocysteine have been implicated as critical factors in the pathogenesis of Alzheimer's disease. The neuroprotective effects and possible mechanism of four structurally similar dibenzocyclooctadiene lignans (namely schisandrin, schisantherin A, schisandrin B and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service