Skip to Content
Merck
All Photos(1)

Key Documents

SCP0249

Sigma-Aldrich

Pancreatic Elastase substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H43N5O8
Molecular Weight:
625.71
UNSPSC Code:
12352200
NACRES:
NA.32

product name

Pancreatic Elastase substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

MeOSuc-Ala-Ala-Pro-Val-4-MNA

Application

Elastase(s) can be assayed and differentiated using chromogenic and fluorogenic Pro-Val peptide substrates such as Glp-Pro-Val-pNA and MeOSuc-Ala-Ala-Pro-Val-4-MNA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Moriuchi et al.
Intensive care medicine, 24(7), 709-715 (1998-08-29)
Oleic acid (OA) can produce a lung injury similar to the adult respiratory distress syndrome (ARDS). Elastase and superoxides are thought to have an effect in ARDS. However, the effect that elastase and superoxide have in OA lung injury is
D Nunley et al.
American journal of respiratory and critical care medicine, 159(1), 258-261 (1999-01-05)
Large numbers of neutrophils with unopposed neutrophil elastase (NE) proteolytic activity are found in lower respiratory tract secretions from most patients with advanced cystic fibrosis (CF). To determine whether antielastase defenses may be overwhelmed in epithelial lining fluid after lung
J V Edwards et al.
The journal of peptide research : official journal of the American Peptide Society, 66(4), 160-168 (2005-09-06)
An assessment of elastase-substrate kinetics and adsorption at the solid-liquid interface of peptide-bound resin was made in an approach to the solid-phase detection of human neutrophil elastase (HNE), which is found in high concentration in chronic wound fluid. N-succinyl-alanine-alanine-proline-valine-p-nitroanilide (suc-Ala-Ala-Pro-Val-pNA)
G Dent et al.
Lung, 173(3), 165-175 (1995-01-01)
Elastase and antielastase activities were measured in bronchoalveolar lavage fluid (BALF) and their relationship to bronchoalveolar lavage (BAL) neutrophil numbers was assessed in order to determine whether the elevated BAL neutrophil count can predict a shift in the elastase/antielastase balance.
Pier Carlo Braga et al.
Pharmacology, 77(3), 150-154 (2006-07-11)
Erdosteine is a mucolytic drug whose metabolization gives rise to an active metabolite with an SH group (Met I), which reduces neutrophil release of reactive oxygen species and the peroxynitrite generated by the reaction of superoxide anion with nitric oxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service