Skip to Content
Merck
All Photos(3)

Key Documents

M9692

Sigma-Aldrich

Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody, Mouse monoclonal

clone MAPK-YT, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MAPK-YT, monoclonal

form

buffered aqueous solution

mol wt

antigen ERK-1 44 kDa
antigen ERK-2 42 kDa

species reactivity

human, Caenorhabditis elegans, Xenopus, Drosophila, hamster, rat, bovine, mouse, yeast

packaging

antibody small pack of 25 μL

concentration

1.5-2 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.5-1 μg/mL using a whole cell extract of RAT-1 cells treated with vanadate and H2O2.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Mitogen-activated protein kinase (MAPK) are also known as the extracellular signal-regulated kinase (ERKs), they exist in two isoforms namely, ERK1 (MAPK3) and ERK2 (MAPK1). ERK2 is mapped to human chromosome 22q11.2. ERK1 is located on human chromosome 16p11.2.
Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the MAPK-YT hybridoma produced by the fusion of mouse myeloma cells (NS1) and splenocytes from BALB/c mice immunized with a synthetic phosphorylated peptide containing amino acids corresponding to the phosphorylated form of ERK-activation loop, conjugated to KLH. The isotype is determined by a double diffusion immunoassay using Mouse Monoclonal Antibody Isotyping Reagents, Product Number ISO2.

Specificity

Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) is specific for the active, dually-phosphorylated form of MAP kinase (ERK-1 and ERK-2, 44 kDa and 42 kDa, respectively). Reactivity has been observed with human, bovine, rat, mouse, hamster, Xenopus, Drosophila, Spodoptera frugiperda, C. elegans, and yeast.
The antibody reacts specifically with the diphosphorylated form of MAP kinase (ERK-1 and ERK-2). It does not recognize the non-phosphorylated or the monophosphorylated forms of MAP kinase or the diphosphorylated forms of JNK and p38 MAP kinase. The epitope recognized by the antibody contains the phosphorylated threonine and tyrosine residues within the regulatory site of active MAP kinase.

Immunogen

synthetic peptide HTGFLpTEpYVAT corresponding to the phosphorylated form of the ERK-activation loop.

Application

The antibody may be used for immunoblotting applications at a working concentration of 0.5-1.0 μg/mL in cultured cells as well as tissue extracts. A working dilution of 1:80-1:100 is suitable for immunohistochemistry (formalin and formaldehyde-fixed sections). The antibody may also be used for immunocytochemistry, immunoprecipitation, indirect ELISA and protein microarray.
Monoclonal Anti-MAP Kinase, activated (Diphosphorylated ERK-1&2) antibody produced in mouse has been used in detection of
  • ERK1/2 in cardiac myocytes using immunofluorescence microscopy
  • ERK1/2 in human eosinophils using western blotting
  • ERK in human bronchial epithelial cells using protein array
  • ERK proteins in human liver cell lines using immunofluorescence assay and western blot analysis
  • ERK1/2 in human embryonic kidney cells by immunoprecipitation

Biochem/physiol Actions

Mitogen-activated protein kinase (MAPK) superfamily of enzymes is involved in widespread signalling pathways. Members of this family include the ERK1/2 (extracellular signal-regulated protein kinase, also termed p42/p44 MAPK), JNK and p38 MAPK subfamilies. These are the terminal enzymes in a signalling cascade where each kinase phosphorylates and activates the next member in the sequence. Phosphorylation of both tyrosine and threonine is essential for the full activation of all MAPKs. Several kinases participate in activation of the ERK cascade. This cascade is initiated by the small G protein Ras, which upon stimulation causes activation Raf1 kinase. Raf1 continues the transmission by activating MEK. Activated MEK appears to be the only kinase capable of specifically phosphorylating and activating ERK. ERK appears to be an important regulatory molecule, which by can phosphorylate regulatory targets in the cytosol (phospholipase A2, PLA2), translocated into and phosphorylate substrates in the nucleus (ELK1). The activation of ERK cascade mediates and regulates the signal transduction pathways in response to stress, mitogenic signals and is important in development and differentiation, learning, memory and survival.
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1 and 2) is specific for the active, dually-phosphorylated form of MAP kinase (ERK-1 and ERK-2, 44 kDa and 42 kDa, respectively). The epitope recognized by the antibody contains the phosphorylated threonine and tyrosine residues within the regulatory site of active MAP kinase (Thr183 and Tyr185 in ERK-2). It does not recognize the non-phosphorylated or the monophosphorylated forms of the MAP kinase molecule or the diphosphorylated form of Jun-kinase (JNK) and p38 MAP kinase.
Signaling pathways mediated by MAPK are associated in the pathogenesis of neurodegenerative disorders and cancer. Deletion in the ERK1 gene locus is associated with DiGeorge syndrome (DGS) and velocardiofacial syndrome (VCFS) and congenital heart defects. Rearrangements in MAPK3 gene locus may contribute in the pathogenesis of autism spectrum disorders and schizophrenia.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots at -20 °C. Repeated freezing and thawing, or storage in frost free freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ERK1 and ERK2 map kinases: specific roles or functional redundancy?
Pouyssegur J and Lenormand P
Frontiers in Cell and Developmental Biology, 4, 53-53 (2016)
Pathological roles of MAPK signaling pathways in human diseases.
Kim EK and Choi EJ
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease, 1802(4), 396-405 (2010)
Long non-coding RNA Igf2as controls hepatocellular carcinoma progression through the ERK/MAPK signaling pathway.
Bao H, et al.
Oncology Letters, 14(3), 2831-2837 (2017)
Congenital heart defects in a novel recurrent 22q11. 2 deletion harboring the genes CRKL and MAPK1.
Breckpot J, et al.
American Journal of Medical Genetics, 158(3), 574-580 (2012)
The 16p11. 2 locus modulates brain structures common to autism, schizophrenia and obesity.
Maillard AM, et al.
Molecular Psychiatry, 20(1), 140-140 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service