Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300161

Sigma-Aldrich

Monoclonal Anti-Ractopamine antibody produced in mouse

clone Rac1, ascites fluid

Synonym(s):

N/A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

Rac1, monoclonal

species reactivity

human

technique(s)

direct ELISA: 1:10,000
western blot: 1:500-1:2,000

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

Rac-OVA.
Mouse monoclonal antibody raised against Ractopamine

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cansu Karabiyik et al.
Brain pathology (Zurich, Switzerland), 28(4), 569-580 (2017-09-30)
The Rho GTPase Rac1 is a multifunctional protein involved in distinct pathways ranging from development to pathology. The aim of the present study was to unravel the contribution of neuronal Rac1 in regulating the response to brain injury induced by
Diane Chan et al.
The Journal of biological chemistry, 286(18), 16140-16149 (2011-04-02)
Mutations in leucine-rich repeat kinase 2 (LRRK2) are currently the most common genetic cause of familial late-onset Parkinson disease, which is clinically indistinguishable from idiopathic disease. The most common pathological mutation in LRRK2, G2019S LRRK2, is known to cause neurite
Wenjing Ruan et al.
Molecular biology of the cell, 19(8), 3192-3202 (2008-05-30)
Members of the tumor necrosis factor receptor (TNFR) superfamily control cell fate determination, including cell death and differentiation. Fas (CD95) is the prototypical "death receptor" of the TNFR superfamily and signals apoptosis through well established pathways. In the adult nervous
Y R Juste et al.
Methods in molecular biology (Clifton, N.J.), 1880, 703-727 (2019-01-06)
Chaperone-mediated autophagy (CMA) is a selective type of autophagy whereby a specific subset of intracellular proteins is targeted to the lysosome for degradation. These proteins are identified by a chaperone that targets them to lysosomes. There, they are translocated into
Hongman Song et al.
Investigative ophthalmology & visual science, 55(4), 2659-2668 (2014-03-22)
Dominant-active RAC1 rescues photoreceptor structure in Drosophila rhodopsin-null mutants, indicating an important role in morphogenesis. This report assesses the morphogenetic effect of activated RAC1 during mammalian rod photoreceptor development using transgenic mice that express constitutively active (CA) RAC1. Transgenic mice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service