Skip to Content
Merck
All Photos(1)

Key Documents

61678

Sigma-Aldrich

NIR-628

suitable for fluorescence, ≥98.0% (HPCE)

Synonym(s):

2,10-Bis(3-aminopropoxy)dibenzo[a,j]perylene-8,16-dione, 3,3′-[(8,16-Dihydro-8,16-dioxodibenzo[a,j]perylen-2,10-diyl)dioxy]dipropylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H28N2O4
CAS Number:
Molecular Weight:
528.60
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98.0% (HPCE)

form

solid

mp

186-190 °C

solubility

acetonitrile: 0.5%, clear

fluorescence

λex 610 nm; λem 658 nm in acetonitrile

suitability

suitable for fluorescence

SMILES string

NCCCOc1ccc-2c(c1)C(=O)c3cccc4c5-c6ccc(OCCCN)cc6C(=O)c7cccc(c-2c34)c57

InChI

1S/C34H28N2O4/c35-13-3-15-39-19-10-12-22-27(17-19)33(37)25-7-1-5-23-29-21-11-9-20(40-16-4-14-36)18-28(21)34(38)26-8-2-6-24(32(26)29)30(22)31(23)25/h1-2,5-12,17-18H,3-4,13-16,35-36H2

InChI key

GLYICBWZRQDSNT-UHFFFAOYSA-N

Application

NIR-628, a near-infrared (NIR) fluorescence dye, is used in applications where the intrinsic fluorescence of biomolecules poses a problem in the assay. Near-infrared (NIR) fluorescence (650–900 nm) detection avoids the natural background fluorescence interference of biomolecules, providing a high contrast between target and background tissues.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabian Niedermair et al.
Inorganic chemistry, 49(20), 9333-9342 (2010-09-16)
A series of π-extended phosphorescent palladium(II) and platinum(II) porphyrin complexes were synthesized, in which additional benzene rings are fused radially onto at least one of the four peripheral benzo groups. The photophysical properties of the metalloporphyrins palladium(II)-meso-tetra-(4-fluorophenyl)mononaphthotribenzoporphyrin (Pd1NF), cis-palladium(II)-meso-tetra-(4-fluorophenyl)dibenzodinaphthoporphyrin (Pd2NF)
Anne Techen et al.
Journal of colloid and interface science, 377(1), 251-261 (2012-04-24)
The well-known cationic surfactant hexadecyltrimethylammonium bromide (CTAB) was used as a model carrier to study drug-carrier interactions with fluorescence probes (5-hexadecanoylaminofluorescein (HAF) and 2,10-bis-(3-aminopropyloxy)dibenzo[a,j]perylene-8,16-dione (NIR 628) by applying ensemble as well as single molecule fluorescence techniques. The impact of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service