Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501762

Sigma-Aldrich

Anti-HSL antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Hormone sensitive lipase, LIPE, LIPS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 116 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIPE(3991)

General description

Human hormone-sensitive lipase (HSL) gene, containing 9 exons and spanning ~11kB, is mapped to chromosome 19q13.3. HSL is majorly expressed in adipose and steroidogenic tissues, but expressed at low levels in heart and skeletal muscle, macrophages, and islets. Functional HSL exists as a homodimer with high hydrolytic activity. HSL is characterized with a C-terminal catalytic domain, regulatory module and an N-terminal domain responsible for protein–protein and protein–lipid interactions.

Immunogen

The antiserum was produced against synthesized peptide derived from human HSL.

Immunogen Range: 520-569

Biochem/physiol Actions

Hormone-sensitive lipase (HSL) is the major source of energy for most of the tissues. It plays a vital role in hydrolysis of triacylglycerol stored in adipose tissue and possibly steroidogenesis. HSL activity is regulated through reversible phosphorylation by catecholamines and insulin. HSL, also hydrolyzes cholesterol esters in the adrenals, ovaries, testes, and macrophages and plays an essential role in spermatogenesis. HSL mRNA expressed in islets might be implicated in glucose-induced insulin secretion. Abnormality in the protein expression results in obesity, atherosclerosis and type II diabetes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen J Yeaman
The Biochemical journal, 379(Pt 1), 11-22 (2004-01-17)
Although described initially as an intracellular adipocyte-specific triacylglycerol lipase, it is now clear that HSL (hormone-sensitive lipase) is expressed in multiple tissues and plays a number of roles in lipid metabolism, including that of a neutral cholesteryl ester hydrolase. The
Fredric B Kraemer et al.
Journal of lipid research, 43(10), 1585-1594 (2002-10-05)
Hormone-sensitive lipase (HSL) is an intracellular neutral lipase that is capable of hydrolyzing triacylglycerols, diacylglycerols, monoacylglycerols, and cholesteryl esters, as well as other lipid and water soluble substrates. HSL activity is regulated post-translationally by phosphorylation and also by pretranslational mechanisms.
Wucai Yang et al.
Scientific reports, 7(1), 15080-15080 (2017-11-10)
Melatonin (N-acetyl-5-methoxytryptamine) is a derivative of tryptophan which is produced and secreted mainly by the pineal gland and regulates a variety of important central and peripheral actions. To examine the potential effects of melatonin on the proliferation and differentiation of
C Holm et al.
Science (New York, N.Y.), 241(4872), 1503-1506 (1988-09-16)
Hormone-sensitive lipase, a key enzyme in fatty acid mobilization, overall energy homeostasis, and possibly steroidogenesis, is acutely controlled through reversible phosphorylation by catecholamines and insulin. The 757-amino acid sequence predicted from a cloned rat adipocyte complementary DNA showed no homology
J Osuga et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(2), 787-792 (2000-01-19)
Hormone-sensitive lipase (HSL) is known to mediate the hydrolysis not only of triacylglycerol stored in adipose tissue but also of cholesterol esters in the adrenals, ovaries, testes, and macrophages. To elucidate its precise role in the development of obesity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service