Skip to Content
Merck
All Photos(2)

Documents

SAB4500662

Sigma-Aldrich

Anti-Aggrecan (Cleaved-Asp369), N-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CSPCP, CSPG1, aggrecan core protein, cartilage-specific proteoglycan core protein, chondroitin sulfate proteoglycan core protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 44 kDa

species reactivity

human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Asp369)

Gene Information

human ... ACAN(176)

Related Categories

General description

Anti-Aggrecan (Cleaved-Asp369) Antibody detects endogenous levels of fragment of activated Aggrecan (Cleaved-Asp369) protein.
Aggrecan is a large aggregating chondroitin sulfate proteoglycan of the human cartilage. It is encoded by AGC1 (aggrecan) gene. It is located on human chromosome 15q26.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human Aggrecan.

Immunogen Range: 320-369

Application

Anti-Aggrecan (Cleaved-Asp369), N-Terminal antibody has been used in western blotting and immunostaining.

Biochem/physiol Actions

Mutations in aggrecan results in autosomal dominant short stature with accelerated skeletal maturation. Mutation in the variable repeat region of the aggrecan gene (AGC1) leads to spondyloepiphyseal dysplasia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gene expression profiles of early chondrogenic markers in dedifferentiated fat cells stimulated by bone morphogenetic protein 4 under monolayer and spheroid culture conditions in vitro
Eiko A, et al.
Orthodontic Waves, 75(4), 97-104 (2016)
Clinical Characterization of Patients With Autosomal Dominant Short Stature due to Aggrecan Mutations.
Gkourogianni A, et al.
The Journal of Clinical Endocrinology and Metabolism, 102(2), 460-469 (2017)
Feng-Lai Yuan et al.
Cell stress & chaperones, 21(1), 97-104 (2015-09-20)
Acidic conditions are present in degenerated intervertebral discs and are believed to be responsible for matrix breakdown. Acid-sensing ion channel 1a (ASIC1a) is expressed in endplate chondrocytes, and its activation is associated with endplate chondrocyte apoptosis. However, the precise role
A Mutation in the Variable Repeat Region of the Aggrecan Gene (AGC1) Causes a Form of Spondyloepiphyseal Dysplasia Associated with Severe, Premature Osteoarthritis
Lindsay G, et al.
American Journal of Human Genetics, 77(3), 484-490 (2005)
Complete coding sequence and deduced primary structure of the human cartilage large aggregating proteoglycan, aggrecan. Human-specific repeats, and additional alternatively spliced forms.
Doege K J, et al.
The Journal of Biological Chemistry, 266(2), 894-902 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service