Skip to Content
Merck
All Photos(2)

Key Documents

SAB4301075

Sigma-Aldrich

Anti-DGAT1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ARAT, ARGP1, DGAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

rat, mouse, human

concentration

2.8 mg/mL

technique(s)

immunohistochemistry: 1:50- 1:200
western blot: 1:500-1:2000 (Cell Lysate)

isotype

IgG

accession no.

NP_036211.2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DGAT1(8694)

General description

Diacylglycerol-acyltransferase 1 (DGAT1), a transmembrane protein, is a member of the membrane-bound O-acyltransferase superfamily. It is seen in the endoplasmic reticulum of several cells. DGAT1 is ubiquitously expressed in humans. A high level of mRNA is seen in the small intestine. It has three transmembrane domains. The DGAT1 gene is mapped to human chromosome 8q24.3.

Specificity

The antibody detects endogenous levels of total DGAT1 protein.

Immunogen

Synthetic peptide corresponding to a region derived from internal residues of human diacylglycerol O-acyltransferase 1

Application

Anti-DGAT1 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Diacylglycerol-acyltransferase 1 (DGAT1) converts diacylglycerol and fatty acyl-CoA to triacylglycerol. It participates in energy storage. DGAT1 plays a key role in cancer progression. Activity of DGAT1 is essential for the physiologic absorption of vitamin D. Absence of DGAT1 activity is linked with body weight and adiposity reduction, and increased insulin sensitivity. Aberrations in the DGAT1 gene are associated with congenital diarrheal disorder (CDD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping He et al.
BMC cancer, 21(1), 252-252 (2021-03-23)
Diacylglycerol-acyltransferase 1 (DGAT1) plays an important role in the energy storage and is involved in cancer progression. A growing number of evidences showed that elevated expression of DGAT1 in cancer tissue indicated a poor outcome in cancer patients. However, the
A Mercadé et al.
Journal of animal breeding and genetics = Zeitschrift fur Tierzuchtung und Zuchtungsbiologie, 122(3), 161-164 (2005-09-01)
DGAT1 is a microsomal enzyme that catalyses the final step in triglycerides synthesis. DGAT1-deficient mice are viable, lean, fertile and resistant to diet induced obesity. We have previously identified a quantitative trait loci (QTL) on chromosome 4 that affects fatty
Aditi Gupta et al.
European journal of medical genetics, 63(4), 103817-103817 (2019-11-30)
DGAT1, a gene encoding a protein involved in lipid metabolism, has been recently implicated in causing a rare nutritional and digestive disease presenting as Congenital Diarrheal Disorder (CDD). Genetic causes of malnutrition can be classified as metabolic disorders, caused by
Chunyan Feng et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1863(8), 834-843 (2018-04-21)
Levels of polyunsaturated phosphatidylcholine (PC) influence plasma membrane structure and function. Phosphatidylcholine (PC) is synthesized de novo in the Kennedy pathway and then undergoes extensive deacylation/reacylation remodeling via Lands' cycle (non-Kennedy pathway). The reacylation is catalyzed by lysophosphatidylcholine acyltransferase (LPCAT)
Justin Chun et al.
Journal of the American Society of Nephrology : JASN, 33(5), 889-907 (2022-03-03)
Two variants in the gene encoding apolipoprotein L1 (APOL1) that are highly associated with African ancestry are major contributors to the large racial disparity in rates of human kidney disease. We previously demonstrated that recruitment of APOL1 risk variants G1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service