Skip to Content
Merck

Skip To

A1978

Anti-β-Actin (ACTB) Antibody

mouse monoclonal, AC-15

Synonym(s):

Monoclonal Anti-β-Actin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
AC-15, monoclonal
Application:
ARR, ICC, IF, IHC (f), WB
Citations:
4100
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture

biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AC-15, monoclonal

form

buffered aqueous solution

mol wt

antigen 42 kDa

species reactivity

sheep, carp, feline, chicken, rat, mouse, Hirudo medicinalis, rabbit, canine, pig, human, bovine, guinea pig

should not react with

Dictyostelium discoideum

technique(s)

immunocytochemistry: 10-40 μg/mL using human foreskin fibroblasts, immunohistochemistry (frozen sections): suitable, indirect immunofluorescence: suitable, microarray: suitable, western blot: 0.5-1 μg/mL using cell extract of human foreskin fibroblasts or chicken fibroblasts.

isotype

IgG1

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTB(60)
mouse ... Actb(11461)
rat ... Actb(81822)

General description

In staining of chicken gizzard ultrathin tissue cryosections, the antibody labels the dense bodies and longitudinal channels linking consecutive dense bodies that are also occupied by desmin and the membrane-associated dense plaque. It does not stain adult cardiac and skeletal muscles except for traces due to contaminations of the sample with non-muscle cells, or if embryonic tissue is being used.
The ACTB (β-actin) gene is mapped to human chromosome 7p22.1. β-Actin is the most abundant protein localized to the cytoplasm. ACTB is expressed ubiquitously. Actin is one of the most conserved eukaryotic proteins, it is expressed in mammals and birds. Four of the actin isoforms represent the differentiation markers of muscle tissues and two are found in almost all cells. Anti-β-Actin antibody, Mouse Monoclonal (mouse IgG1 isotype) is derived from the AC-15 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a slightly modified synthetic b-cytoplasmic actin N-terminal peptide Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys, conjugated to KLH.
In staining of chicken gizzard ultrathin tissue cryosections, the antibody labels the dense bodies and longitudinal channels linking consecutive dense bodies that are also occupied by desmin and the membrane-associated dense plaque. It does not stain adult cardiac and skeletal muscles except for traces due to contaminations of the sample with non-muscle cells, or if embryonic tissue is being used.

Immunogen

slightly modified β-cytoplasmic actin N-terminal peptide, Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys, conjugated to KLH.

Application

Monoclonal anti-beta-actin antibody can be used for microarray, indirect immunofluorescence, and immunohistochemical analyses. Furthermore, the product has been used for immunocytochemistry at 10-40 μg/mL using human foreskin fibroblasts. The antibody has also been used for western blot at 0.5-1 μg/mL using cell extract of human foreskin fibroblasts or chicken fibroblasts.
The antibody can be used for staining of acetone-fixed frozen sections, EM preparations, and microinjection experiments. B5, ethanol, methacam, or Bouin′s solutions can be used as fixatives. The epitope recognized by the antibody is resistant to formalin-fixation and paraffin-embedding.
Monoclonal mouse anti-actin antibody was used as a loading control for western blot analysis of immunoprecipitated proteins from rat dorsal root ganglion cocultures.
Monoclonal mouse anti-actin was used as a loading control for western blot analysis of rat liver protein lysates.
The actin in cells of various species and tissue origin is very similar in their immunological and physical properties. As a consequence, it has been difficult to produce potent antisera to this protein. Therefore the availability of monoclonal antibodies to β-actin provides a specific and useful tool in studying the intracellular distribution of β-actin and the static and dynamic aspects of the cytoskeleton.
Monoclonal anti-beta-actin antibody can be used for microarray, indirect immunofluorescence, and immunohistochemical analyses. Furthermore, the product has been used for immunocytochemistry at 10-40 μg/mL using human foreskin fibroblasts. The antibody has also been used for western blot at 0.5-1 μg/mL using cell extract of human foreskin fibroblasts or chicken fibroblasts.
The antibody can be used for staining of acetone-fixed frozen sections, EM preparations, and microinjection experiments. B5, ethanol, methacam, or Bouin′s solutions can be used as fixatives. The epitope recognized by the antibody is resistant to formalin-fixation and paraffin-embedding.
It has been used in Immunoblot analysis.

Biochem/physiol Actions

Anti-β-Actin antibody, Mouse Monoclonal 1, 2 recognizes an epitope located on the N-terminal end of the β-isoform of actin.
The two major cytoskeletal proteins implicated in cell motility are actin and myosin. Actin and myosin are constituents of many cell types and are involved in myriad of cellular process including locomotion, secretion, cytoplasmic streaming, phagocytosis, and cytokinesis.
ACTB (β-actin) is crucial for organogenesis, especially for the development of brain, kidney and heart. It is responsible for maintaining cell proliferation, migration and shape. Actin protein is essential for the formation of mature platelets. β−Actin plays a key role in embryonic development. Upregulation of β-actin stimulates membrane protrusions and helps in cellular motility. The encoded protein β-Actin mediates fibroblast migration in mouse and thereby control connective tissue tension.
Actin is a cytoskeletal protein that regulates cell motility, secretion, phagocytosis and cytokinesis. The NH2-terminal of actin may function as an antigen. This terminal may also modulate actin interactions and may associate with proteins such as myosin.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Storage and Stability: For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots at -20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Other Notes

To view an Actin antibody selection guide, please visit www.sigmaaldrich.com/actin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F3022A2228WH0000060M1
description

clone AC-15, purified from hybridoma cell culture

description

clone AC-15, purified from hybridoma cell culture

description

clone AC-74, purified immunoglobulin, buffered aqueous solution

description

clone 3G4-F9, purified immunoglobulin, buffered aqueous solution

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

FITC conjugate

conjugate

unconjugated

conjugate

unconjugated

species reactivity

sheep, carp, feline, chicken, rat, mouse, Hirudo medicinalis, rabbit, canine, pig, human, bovine, guinea pig

species reactivity

sheep, carp, feline, chicken, rat, mouse, Hirudo medicinalis, rabbit, canine, pig, human, bovine, guinea pig

species reactivity

rabbit, sheep, cat, guinea pig, bovine, mouse, chicken, wide range, Hirudo medicinalis, canine, Drosophila, carp, pig, rat, human

species reactivity

human, rat, mouse

Gene Information

human ... ACTB(60)
mouse ... Actb(11461)
rat ... Actb(81822)

Gene Information

human ... ACTB(60)
mouse ... Actb(11461)
rat ... Actb(81822)

Gene Information

human ... ACTB(60)
mouse ... Actb(11461)
rat ... Actb(81822)

Gene Information

human ... ACTB(60)

clone

AC-15, monoclonal

clone

AC-15, monoclonal

clone

AC-74, monoclonal

clone

3G4-F9, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

Active Filters

  1. 1 Ratings-only review