Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200779

Sigma-Aldrich

Anti-NRAS Antibody

enhanced validation

rabbit polyclonal

Synonym(s):

GTPase NRas, Transforming protein N-Ras

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-NRAS (C-terminal) antibody produced in rabbit, IgG fraction of antiserum

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt ~21 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 1:2,000-1:4,000 using human HEK-293T cells over-expressing NRAS protein

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NRAS(4893)

General description

Transforming protein N-Ras (NRAS), also known as GTPase NRas or Neuroblastoma RAS Viral (V-Ras) Oncogene Homolog, is a member of the Ras protein family and are low molecular-weight GTPases. NRAS is mapped to human chromosome location 1p13.2 and shows ubiquitous expression. Ras proteins have highly homologous primary amino acid sequence and differ in their C-terminal region termed as hypervariable region (HVR).

Specificity

Anti-NRAS (C-terminal) antibody specifically recognizes human NRAS and does not cross-react with HRAS or KRAS.

Immunogen

Synthetic peptide from the internal region of human NRAS protein, conjugated to KLH

Application

Anti-NRAS (C-terminal) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Neuroblastoma RAS Viral (V-Ras) Oncogene Homolog (NRAS) was the first melanoma oncogene to be identified. Oncogenic NRAS mutations are single base substitutions (most commonly affecting residues G12, G13, or Q61) that lead to the stabilization of GTP binding and constitutive activation of RAS and downstream signaling cascades. Abnormal NRAS activity stimulates several signaling pathways, including mitogen-activated protein kinase (MAPK/ERK), serine/threonine-protein kinase (RAFs) (ARAF, BRAF, and CRAF), phosphatidylinositol 3-kinase (PI3K) and the RAS-like protein (RAL) guanine nucleotide exchange factors (GEFs) signaling pathways. This leads to uncontrolled cell proliferation, resistance to apoptosis and thus cancer therapy potential target. NRAS mutations are present in various cancers, including melanomas, acute myeloid leukemia, colon, thyroid and lung cancers. Mutations are also implicated in hematologic malignancies, including acute lymphocytic leukemia, myelodysplastic syndrome, multiple myeloma and chronic myelomonocytic leukemia.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional specificity of ras isoforms: so similar but so different
Castellano, Esther and Santos, Eugenio
Genes & Cancer, 2(3), 216-231 (2011)
Eva Muñoz-Couselo et al.
OncoTargets and therapy, 10, 3941-3947 (2017-09-02)
Melanoma is one of the most common cutaneous cancers worldwide. Activating mutations in RAS oncogenes are found in a third of all human cancers and NRAS mutations are found in 15%-20% of melanomas. The NRAS-mutant subset of melanoma is more
Genetic alterations in RAS-regulated pathway in acral lentiginous melanoma
Puig-Butille JA, et al.
Experimental Dermatology, 22(2), 148-150 (2013)
Molecular pathways: targeting NRAS in melanoma and acute myelogenous leukemia
Johnson DB, et al.
Clinical Cancer Research, 20(16), 4186-4192 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service