Skip to Content
Merck
All Photos(9)

Key Documents

ZRB003845

Sigma-Aldrich

Anti-KRas Antibody, clone 2J13, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

GTPase KRas, K-Ras 2, Ki-Ras, c-K-ras, c-Ki-ras

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2J13, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 21.66 kDa
observed mol wt ~21 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

gene knockdown: suitable using 1:1,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:400
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2J13 is a ZooMAb rabbit recombinant monoclonal antibody that detects human KRas.

Immunogen

GST/His-tagged full-length recombinant human KRas.

Application

Anti-KRas, clone 2J13, ZooMAb, Cat. No. ZRB003845, is a recombinant Rabbit Monoclonal antibody that detects human KRas and has been tested for use in Immunohistochemistry (Paraffin), RNAi/siRNA/Gene Knockdown, and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:400 dilution from a representative lot detected KRas in human kidney tissue sections.

RNAi/siRNA/Gene Knockdown Analysis: A 1:1,000 dilution from a representative lot detected KRas in lysates from wild type HeLa cells, but not in lysate from KRAS knockout HeLa cells.

Target description

GTPase KRas (UniProt P01116; also known as c-K-Ras, c-Ki-Ras, K-Ras 2, Ki-Ras) is encoded by the KRAS (also known as KRAS2, RASK2) gene (Gene ID 3845) in human. Ras proteins are small GTPases, which control signaling pathways for growth, proliferation, and differentiation. Mammalian genomes harbor three Ras genes (HRAS, NRAS, and KRAS) that encode four GTPases, two of which (K-Ras4A and K-Ras4B) are transcribed from the KRAS gene via two alternative fourth exons. Ras proteins share nearly identical sequence homology in sequence encoded by exons 1-3 (a.a. 1-165), while diverge in their C-terminal 24-a.a. hypervariable region (HVR) encoded by exon 4. The first 165 amino acids form the catalytic G domain that mediates GDP/GTP binding and association with effectors, exchange factors, and GTPase-activating proteins (GAPs), while the C-terminal HVR is needed for Ras membrane localization, a process necessary for Ras function. In addition to the removal of initiator methionine (Met1), Ras proteins are post-translationally modified by farnesylation of Cys186, followed by proteolytical cleavage their C-terminal end three amino acids (aa 187-189) and carboxyl methylation of the newly formed C-terminal prenylcysteine. H-Ras, N-Ras, and K-Ras4A are further palmitoylated on cysteine residue(s) near the C-terminus (Cys181/Cys184 in H-Ras, Cys181 in N- and K-Ras), while K-Ras4B lacks a palmitoylation site. Palmitoylation is required for targeting H- and N-Ras to the plasma membrane (PM), while K-Ras4A and K-Ras4B contain a series of lysines that constitute a polybasic region (PBR) capable of interacting with the inner leaflet of the PM through an electrostatic interaction. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service