Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500346

Sigma-Aldrich

Anti-ACE2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ACE2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACE2(59272)

General description

Angiotensin-converting enzyme-2 (ACE2), is a membrane-associated and secreted enzyme, encoded by the gene mapped to human chromosome Xp22.2. Ace 2 is a human homolog of ACE and is expressed mainly on vascular endothelium of heart, kidney, and testis. Ace2 structure comprises a N-terminal PD and a C-terminal collectrin-like domain (CLD).

Immunogen

ACE2 antibody was raised against a synthetic peptide corresponding to amino acids near the center of human ACE2.

Biochem/physiol Actions

Angiotensin-converting enzyme-2 (ACE2) catalyzes the conversion of Ang I to Ang1-9. It might also be involved in maintaining the balance of local renin-angiotensin system (RAS) in heart and kidney. Ace 2 acts as a functional receptor for spike glycoprotein of severe acute respiratory syndrome-coronavirus (SARS-CoV) and the new coronavirus (SARS-CoV-2), which is a causative agent for coronavirus disease 2019 (COVID-19). Downregulated expression of Ace 2 causes cardiovascular diseases.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500346.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clara Husser et al.
Microorganisms, 12(3) (2024-03-28)
While having already killed more than 7 million of people worldwide in 4 years, SARS-CoV-2, the etiological agent of COVID-19, is still circulating and evolving. Understanding the pathogenesis of the virus is of capital importance. It was shown that in
Lin Wang et al.
Theranostics, 11(2), 649-664 (2021-01-05)
The novel severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has caused a worldwide epidemic of the lethal respiratory coronavirus disease (COVID-19), necessitating urgent development of specific and effective therapeutic tools. Among several therapeutic targets of coronaviruses, the spike protein is
Shine Varghese Jancy et al.
Biological procedures online, 25(1), 22-22 (2023-07-27)
The entry of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) into the host cell is mediated through the binding of the SARS-CoV-2 Spike protein via the receptor binding domain (RBD) to human angiotensin-converting enzyme 2 (hACE2). Identifying compounds that inhibit
Elisa Avolio et al.
Clinical science (London, England : 1979), 135(24), 2667-2689 (2021-11-23)
The severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) causes a broad range of clinical responses including prominent microvascular damage. The capacity of SARS-CoV-2 to infect vascular cells is still debated. Additionally, the SARS-CoV-2 Spike (S) protein may act as a
ACE2 is on the X chromosome: could this explain COVID-19 gender differences?
Esther Culebras et al.
European heart journal (2020-06-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service