Skip to Content
Merck
All Photos(1)

Key Documents

07-434

Sigma-Aldrich

Anti-BRCA1 Antibody

serum, Upstate®

Synonym(s):

Anti-BRCAI, Anti-BRCC1, Anti-BROVCA1, Anti-FANCS, Anti-IRIS, Anti-PNCA4, Anti-PPP1R53, Anti-PSCP, Anti-RNF53

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BRCA1(672)

General description

Two major susceptibility genes for breast cancer, BRCA1 and BRCA2, have been identified. Both genes are considered to be tumor-suppressor genes. Mutations within BRCA1 and BRCA2 are responsible for most familial breast cancer cases. Functional analyses of the BRCA1 and BRCA2 gene products have established their dual participation in transcription regulation and DNA damage repair. BRCA1 affects cell cycle regulation and loss of BRCA1 function due to decreased expression leads to cell cycle arrest, through p53 and p21 genes. In vivo, BRCA1 and BRCA2 are expressed at maximal levels in rapidly proliferating cells. This feature is consistent with in vitro observations that BRCA1 and BRCA2 are expressed in a cell cycle-dependent manner. During mammary gland development, the expression of BRCA1 and BRCA2 is induced in rapidly proliferating cellular compartments.

Specificity

Recognizes human BRCA1.

Immunogen

GST-tagged fusion protein corresponding to amino acids 1301-1863 of human BRCA1.

Application

Detect BRCA1 with Anti-BRCA1 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in IHC, IP & WB. This BRCA1 antibody has been tested in human samples.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated by immunoblot in HeLa nuclear extract

Target description

220 kDa

Physical form

Serum containing 0.035% sodium azide and 30% glycerol. Liquid at -20°C.
Unpurified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ligand-dependent differences in estrogen receptor beta-interacting proteins identified in lung adenocarcinoma cells corresponds to estrogenic responses.
Ivanova, M; Abner, S; Pierce, W; Klinge, C
Proteome Science null
BAL1 and its partner E3 ligase, BBAP, link Poly(ADP-ribose) activation, ubiquitylation, and double-strand DNA repair independent of ATM, MDC1, and RNF8.
Yan, Q; Xu, R; Zhu, L; Cheng, X; Wang, Z; Manis, J; Shipp, MA
Molecular and cellular biology null
Aurélia Noll et al.
Cancer cell international, 16, 53-53 (2016-07-05)
Poly(ADP-ribose) polymerase (PARP) inhibitors have entered the clinics for their promising anticancer effect as adjuvant in chemo- and radiotherapy and as single agent on BRCA-mutated tumours. Poly(ADP-ribose) glycohydrolase (PARG) deficiency was also shown to potentiate the cytotoxicity of genotoxic agents
Nicole L Batenburg et al.
Nucleic acids research, 47(20), 10678-10692 (2019-09-11)
CSB, a member of the SWI2/SNF2 superfamily, has been implicated in evicting histones to promote the DSB pathway choice towards homologous recombination (HR) repair. However, how CSB promotes HR repair remains poorly characterized. Here we demonstrate that CSB interacts with
Qingsheng Yan et al.
Molecular cell, 36(1), 110-120 (2009-10-13)
Although the BBAP E3 ligase and its binding partner BAL are overexpressed in chemotherapy-resistant lymphomas, the role of these proteins in DNA damage responses remains undefined. Because BAL proteins modulate promoter-coupled transcription and contain structural motifs associated with chromatin remodeling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service