Skip to Content
Merck
All Photos(3)

Documents

SAB3500285

Sigma-Aldrich

Anti-LAMP-1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Lysosome associated membrane protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 46 kDa

species reactivity

rat, human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMP1(3916)

General description

Lysosomal-associated membrane protein 1 (LAMP-1) is a type I transmembrane protein with a short cytoplasmic tail containing a lysosome-targeting signal. The gene encoding this protein is localized on human chromosome 13q34.

Immunogen

LAMP-1 antibody was raised against a 15 amino acid peptide from near the center of human LAMP-1.

Application

Anti-LAMP-1 antibody produced in rabbit has been used in Western blotting.

Biochem/physiol Actions

Lysosomal-associated membrane protein 1 (LAMP-1) functions as an intracellular receptor. It is found to be expressed in the cytoplasm of several types of tumor cells and may be involved in tumor invasion. LAMP-1 is essential for perforin trafficking to the lytic granules and motility of these lytic granules. Knockdown of this protein leads to inhibition of cytotoxicity of human natural killer cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500285.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A dynamic network of estrogen receptors
in murine lymphocytes: fine-tuning
the immune response
Andrea E. Schneider
Journal of Leukocyte Biology (2017)
LAMP1/CD107a is required for efficient perforin delivery to lytic granules and NK-cell cytotoxicity.
Krzewski K
Blood (2013)
Monosomy and ring chromosome 13 in a thyroid nodular goiter-do we underestimate its relevance in benign thyroid lesions?
Sendt W
Cancer Genetics (2012)
Expression of the lysosomal-associated membrane protein-1 (LAMP-1) in astrocytomas.
Jensen SS
International Journal of Clinical and Experimental Pathology (2013)
Constitutive expression of a COOH-terminal leucine mutant of lysosome-associated membrane protein-1 causes its exclusive localization in low density intracellular vesicles.
Akasaki K
Journal of Biochemistry (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service