Skip to Content
Merck
All Photos(7)

Key Documents

SAB2702136

Sigma-Aldrich

Monoclonal Anti-BRCA1 antibody produced in mouse

Synonym(s):

PNCA4, PPP1R53, PSCP, RNF53

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6B4
monoclonal

form

liquid

mol wt

220 kDa

species reactivity

mouse, human

concentration

1.36 mg/mL

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 1:500-1:3000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRCA1(672)

General description

The gene BRCA1 is mapped to human chromosome 17q21 and codes for a protein with zinc finger motif and a set of acidic residues. BRCA1 is localized to the nucleus and cytoplasm.

Immunogen

BRCA1 protein fragment expressed in E. coli corresponding to amino acids 341-748.

Application

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000, IHC, IP, ChIP. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

BRCA1-6B4 recognizes full length BRCA1, a 220 kDa nuclear phosphoprotein, and does not recognize the exon 11 splice variant. Mutations in this tumor suppressor gene greatly increase the risk of breast cancer. In a high proportion of breast and ovarian cancer cell lines, BRCA1 aberrantly mislocates to the cytoplasm. Recent studies suggest a role for BRCA1 in DNA double strand break repair because of its association with Rad51. BRCA1 is predominantly expressed in proliferating and differentiating cells and contributes to cellular growth.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The nuclear localization sequences of the BRCA1 protein interact with the importin-α subunit of the nuclear transport signal receptor.
Chen C F, et al.
The Journal of Biological Chemistry, 271(51), 32863-32868 (1996)
Epidemiology of breast cancer.
Key T J, et al.
Lancet Oncology, 2(3), 133-140 (2001)
Location of BRCA1 in human breast and ovarian cancer cells.
Scully R, et al.
Science, 272(5258), 123-123 (1996)
Anuratha Sakthianandeswaren et al.
Cancer discovery, 8(8), 988-1005 (2018-06-09)
ADP-ribosylation is an important posttranslational protein modification that regulates diverse biological processes, controlled by dedicated transferases and hydrolases. Here, we show that frequent deletions (∼30%) of the MACROD2 mono-ADP-ribosylhydrolase locus in human colorectal cancer cause impaired PARP1 transferase activity in
Michal Franek et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 64(11), 669-686 (2016-09-30)
DNA damage response (DDR) in ribosomal genes and mechanisms of DNA repair in embryonic stem cells (ESCs) are less explored nuclear events. DDR in ESCs should be unique due to their high proliferation rate, expression of pluripotency factors, and specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service