Skip to Content
Merck
All Photos(2)

Key Documents

SAB1401420

Sigma-Aldrich

Anti-SYN3 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYN3(8224)

General description

This gene is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. The protein encoded by this gene shares the synapsin family domain model, with domains A, C, and E exhibiting the highest degree of conservation. The protein contains a unique domain J, located between domains C and E. Based on this gene′s localization to 22q12.3, a possible schizophrenia susceptibility locus, and the established neurobiological roles of the synapsins, this family member may represent a candidate gene for schizophrenia. The TIMP3 gene is located within an intron of this gene and is transcribed in the opposite direction. Alternative splicing of this gene results in multiple splice variants that encode different isoforms. (provided by RefSeq)

Immunogen

SYN3 (AAH75065.1, 1 a.a. ~ 580 a.a) full-length human protein.

Sequence
MNFLRRRLSDSSFMANLPNGYMTDLQRPDSSTSSPASPAMERRHPQPLAASFSSPGSSLFSSLSSAMKQAPQATSGLMEPPGPSTPIVQRPRILLVIDDAHTDWSKYFHGKKVNGEIEIRVEQAEFSELNLAAYVTGGCMVDMQVVRNGTKVVSRSFKPDFILVRQHAYSMALGEDYRSLVIGLQYGGLPAVNSLYSVYNFCSKPWVFSQLIKIFHSLGPEKFPLVEQTFFPNHKPMVTAPHFPVVVKLGHAHAGMGKIKVENQLDFQDITSVVAMAKTYATTEAFIDSKYDIRIQKIGSNYKAYMRTSISGNWKANTGSAMLEQVAMTERYRLWVDSCSEMFGGLDICAVKAVHSKDGRDYIIEVMDSSMPLIGEHVEEDRQLMADLVVSKMSQLPMPGGTAPSPLRPWAPQIKSAKSPGQAQLGPQLGQPQPRPPPQGGPRQAQSPQPQRSGSPSQQRLSPQGQQPLSPQSGSPQQQRSPGSPQLSRASSGSSPNQASKPGATLASQPRPPVQGRSTSQQGEESKKPAPPHPHLNKSQSLTNSLSTSDTSQRGTPSEDEAKAETIRNLRKSFASLFSD

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service