Skip to Content
Merck
All Photos(8)

Key Documents

HPA000982

Sigma-Aldrich

Anti-STAT1 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Signal transducer and activator of transcription 1-α/β antibody produced in rabbit, Anti-Transcription factor ISGF-3 components p91/p84 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LLRFSESSREGAITFTWVERSQNGGEPDFHAVEPYTKKELSAVTFPDIIRNYKVMAAENIPENPLKYLYPNIDKDHAFGKYYSRPKEAPEPMELDGPKGTGYIKTELISVSEVHPSRLQTTDNLLPMSPEEFDEVSRIVGSVE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAT1(6772)

Looking for similar products? Visit Product Comparison Guide

General description

STAT1 (signal transducer and activator of transcription 1) is the founding member of the STAT family, which in humans consists of seven members. It contains a Src-homology-2-(SH2) domain, and when activated forms homodimers. This gene is localized to human chromosome locus 2q32.2-q32.3.

Immunogen

Signal transducer and activator of transcription 1-α/β recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

STAT1 (signal transducer and activator of transcription 1) is involved in upregulating genes due to a signal by either type I, type II, or type III interferons. STAT1 may be a target for therapeutic treatment to restore apoptotic mechanisms and sensitivity to chemotherapy. P-STAT1 is a potential predictor of interferon (IFN) response for advanced renal cell carcinoma. Mutations in the gene is associated with death at an early age due to overwhelming viral infection (complete STAT1 deficiency).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78216

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Li et al.
Molecular medicine reports, 9(5), 1929-1934 (2014-03-15)
The present study aimed to evaluate the expression status of Janus kinase (JAK)-Signal Transducer and Activator of Transcription (STAT) in renal cell carcinoma and benign renal tissue, and identify a potential biomarker for interferon (IFN) response prediction. A total of
Elisa Espinet et al.
Cancer discovery, 11(3), 638-659 (2020-10-17)
Pancreatic ductal adenocarcinoma (PDAC) is characterized by extensive desmoplasia, which challenges the molecular analyses of bulk tumor samples. Here we FACS-purified epithelial cells from human PDAC and normal pancreas and derived their genome-wide transcriptome and DNA methylome landscapes. Clustering based
Aya Kawasaki et al.
Arthritis research & therapy, 10(5), R113-R113 (2008-09-23)
Recent studies identified STAT4 (signal transducers and activators of transcription-4) as a susceptibility gene for systemic lupus erythematosus (SLE). STAT1 is encoded adjacently to STAT4 on 2q32.2-q32.3, upregulated in peripheral blood mononuclear cells from SLE patients, and functionally relevant to
Alice Ly et al.
Journal of proteome research, 15(4), 1350-1359 (2016-03-05)
The rd10 mouse is a model of retinitis pigmentosa characterized by the dysfunction of a rod-photoreceptor-specific phosphodiesterase. Compared to the rd1 mouse, retinal degeneration in the rd10 mouse begins later in age with a milder phenotype, making it ideal for
Atsushi Tanaka et al.
PloS one, 15(4), e0229252-e0229252 (2020-04-11)
Proteomic analyses indicate that STAT1 protein (signal transducer and activator of transcription 1 or transcription factor ISGF-3 components p91/p84) is upregulated in some colorectal cancers. This study examined 736 colorectal cancer patients for the expression of STAT1 protein in tissue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service