Skip to Content
Merck
All Photos(2)

Documents

S9318

Sigma-Aldrich

Sandoz 58-035

>98% (HPLC), powder, acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor

Synonym(s):

3-[Decyldimethylsilyl]-N-[2-(4-methylphenyl)-1-phenethyl]propanamide, SA 58-035

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H47NOSi
CAS Number:
Molecular Weight:
465.79
MDL number:
UNSPSC Code:
41121801
PubChem Substance ID:
NACRES:
NA.77

product name

Sandoz 58-035, >98% (HPLC), powder

Quality Level

Assay

>98% (HPLC)

form

powder

color

white

solubility

DMSO: 16 mg/mL
H2O: insoluble

originator

Novartis

storage temp.

2-8°C

SMILES string

CCCCCCCCCC[Si](C)(C)CCC(=O)NC(Cc1ccc(C)cc1)c2ccccc2

InChI

1S/C30H47NOSi/c1-5-6-7-8-9-10-11-15-23-33(3,4)24-22-30(32)31-29(28-16-13-12-14-17-28)25-27-20-18-26(2)19-21-27/h12-14,16-21,29H,5-11,15,22-25H2,1-4H3,(H,31,32)

InChI key

NBYATBIMYLFITE-UHFFFAOYSA-N

Gene Information

human ... SOAT1(6646)
rat ... Soat1(81782)

Application

Sandoz 58-035 was used to induce simultaneous activation of unfolded protein response (UPR) and pattern recognition receptors (PRRs) in mouse peritoneal macrophages.3

Biochem/physiol Actions

Sandoz 58-035 inhibits the accumulation of cholesteryl esters and inhibits the esterification of cholesterol by 95% in arterial smooth muscle cells in culture.1 It does not affect the triglyceride metabolism by the gut.2
Acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masato Furuhashi et al.
Scientific reports, 7(1), 217-217 (2017-03-18)
Cholesterol efflux capacity (CEC) from macrophages, the first step in the reverse cholesterol transport pathway, is inversely associated with residual risk for atherosclerotic cardiovascular disease. Fatty acid-binding protein 4 (FABP4) and FABP5 are expressed in both adipocytes and macrophages and
Markus Trieb et al.
Journal of hepatology, 73(1), 113-120 (2020-02-18)
High-density lipoprotein cholesterol (HDL-C) levels are reduced in patients with chronic liver disease and inversely correlate with disease severity. During acute conditions such as sepsis, HDL-C levels decrease rapidly and HDL particles undergo profound changes in their composition and function.
Irene Schilcher et al.
Scientific reports, 7(1), 12485-12485 (2017-10-04)
Endothelial lipase (EL) is a potent modulator of the structural and functional properties of HDL. Impact of EL on cholesterol efflux capacity (CEC) of serum and isolated HDL is not well understood and apparently contradictory data were published. Here, we
Julia T Stadler et al.
Biomedicines, 9(3) (2021-03-07)
Obesity increases the risk of coronary heart disease, partly due to its strong association with atherogenic dyslipidemia, characterized by high triglycerides and low high-density lipoprotein (HDL) cholesterol levels. Functional impairment of HDL may contribute to the increased cardiovascular mortality, but
Shelley J Edmunds et al.
Scientific reports, 10(1), 18070-18070 (2020-10-24)
Apolipoprotein A-I (ApoA-I) of high-density lipoprotein (HDL) induces glucose uptake by muscle tissues and stimulates pancreatic insulin secretion, and also facilitates cholesterol transport in circulation, and is explored for anti-diabetic and anti-atherosclerotic treatments. As the better alternative to complex protein-lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service