Skip to Content
Merck
All Photos(2)

Key Documents

S8504

Sigma-Aldrich

Sulfatase from Patella vulgata (keyhole limpet)

Type IV, essentially salt-free, lyophilized powder, ≥10 units/mg solid

Synonym(s):

Aryl-sulfatase, Aryl-sulfate sulfohydrolase, Phenolsulfatase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

limpet (Patella vulgata)

Quality Level

type

Type IV

form

essentially salt-free, lyophilized powder

specific activity

≥10 units/mg solid

foreign activity

β-Glucuronidase ≥750 units/mg solid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrocatechol sulfate per hr at pH 5.0 at 37 °C (30 min assay).

substrate

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosuke Yusa
Nature protocols, 8(10), 2061-2078 (2013-09-28)
I report here a detailed protocol for seamless genome editing using the piggyBac transposon in human pluripotent stem cells (hPSCs). Recent advances in custom endonucleases have enabled us to routinely perform genome editing in hPSCs. Conventional approaches use the Cre/loxP
S Saha et al.
Pharmacological research, 65(6), 592-602 (2012-03-01)
Epicatechin is a widely consumed dietary flavonoid and there is substantial evidence that it contributes to the health benefits reported for flavanol-rich cocoa products including dark chocolate. Numerous reports have described the appearance of epicatechin and epicatechin phase-2 conjugates (sulfates
P R Warman et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 45(7), 633-638 (2010-08-31)
The study was undertaken to determine the impact of high-metal composts on the activities of four soil enzymes. High concentrations of metal salts (Cr, Cu, Ni or a Co-Mo-Pb combination) were added to feedstocks during the thermophilic stage of composting.
Fatemeh Moheimani et al.
Atherosclerosis, 223(1), 144-151 (2012-06-05)
People with diabetes have an elevated risk of atherosclerosis. The accumulation of lipid within macrophage cells in the artery wall is believed to arise via the uptake and subsequent processing of modified low-density lipoproteins (LDL) via the endo-lysosomal system. In
Christiane Kehrer et al.
Developmental medicine and child neurology, 53(9), 850-855 (2011-06-29)
Motor deterioration is a key feature in metachromatic leukodystrophy (MLD). The lack of data about its natural course impedes evaluation of therapeutic interventions. This study aimed to provide data about motor decline in MLD. Fifty-nine patients (27 males, 32 females)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service