Skip to Content
Merck
All Photos(1)

Documents

S7896

Sigma-Aldrich

Syringaldazine

indicator for laccase and peroxidase activity, ≥98% (TLC), powder or crystals

Synonym(s):

4-Hydroxy-3,5-dimethoxybenzaldehyde azine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H2(OCH3)2CH=N-]2
CAS Number:
Molecular Weight:
360.36
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

Syringaldazine, indicator for laccase and peroxidase activity

Assay

≥98% (TLC)

form

powder or crystals

mp

209-210 °C (lit.)

solubility

DMF: soluble, slightly hazy

storage temp.

2-8°C

SMILES string

COc1cc(\C=N\N=C\c2cc(OC)c(O)c(OC)c2)cc(OC)c1O

InChI

1S/C18H20N2O6/c1-23-13-5-11(6-14(24-2)17(13)21)9-19-20-10-12-7-15(25-3)18(22)16(8-12)26-4/h5-10,21-22H,1-4H3/b19-9+,20-10+

InChI key

YARKTHNUMGKMGS-LQGKIZFRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Syringaldazine has been used as a substrate to determine the enzymatic activity of the cuticular laccase.

Biochem/physiol Actions

Syringaldazine is a dimer of two molecules of 2,6-dimethoxyphenol linked by an azide bridge. It is a phenolic non-autooxidizable laccase-specific compound, that facilitates the detection of fungal laccase in some varieties of fungi. In addition, it also acts as an indicator for peroxidase activity in fungi.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative estimation of laccase forms in some white-rot fungi using syringaldazine as a substrate
Leonowicz A and Grzywnowicz K
Enzyme and Microbial Technology, 3(1), 55-58 (1981)
Use of syringaldazine for detection of laccase in sporophores of wood rotting fungi
Harkin JM, et al.
Mycologia, 66(3), 469-476 (1974)
Aptitude of Oxidative Enzymes for Treatment of Wastewater Pollutants: A Laccase Perspective
Unuofin JO, et al.
Molecules (Basel), 24(11), 2064-2064 (2019)
Characterization of endogenous and recombinant forms of laccase-2, a multicopper oxidase from the tobacco hornworm, Manduca sexta
Dittmer NT, et al.
Insect Biochemistry and Molecular Biology, 39(9), 596-606 (2009)
Syringaldazine, an effective reagent for detecting laccase and peroxidase in fungi
Harkin JM and Obst JR
Experientia, 29(4), 381-387 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service