Skip to Content
Merck
All Photos(1)

Key Documents

S7648

Sigma-Aldrich

Sera from rat

frozen liquid

Synonym(s):

Rat blood, Rat serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

biological source

rat

Quality Level

sterility

sterile

form

frozen liquid

origin

USA origin

shipped in

dry ice

storage temp.

−20°C

Application

Sera from rat has been used:
  • in food folate analysis to prepare rat serum conjugase to deconjugate folates in extract samples
  • in the preparation of deconjugase for enzyme digestion
  • in ex vivo experiments to analyze rat serum spiked with intravenous iron preparations

Biochem/physiol Actions

Rat serum supports the reaggregation of rat islet cells into tridimensional pseudo islets useful for transplantation studies. It functions as an inhibitor of the non-specific binding of rat antibodies in immunochemical techniques.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

V S Balakrishnan et al.
European journal of clinical investigation, 39(6), 489-496 (2009-04-29)
Intravenous iron is a critical component of anaemia management. However, currently available preparations have been associated with the release of free iron, a promoter of bacterial growth and oxidative stress. We determined the molecular weight, dialysability and capacity for free
Johanna E Camara et al.
Analytical and bioanalytical chemistry, 405(13), 4561-4568 (2013-01-29)
The National Institute of Standards and Technology (NIST) is developing a wide variety of Standard Reference Materials (SRMs) to support measurements of vitamins and other nutrients in foods. Previously, NIST has provided SRMs with values assigned for the folate vitamer
L Lione et al.
Bollettino della Societa italiana di biologia sperimentale, 77(1-3), 13-19 (2001-08-22)
Rat islet cells in culture are able to form tridimensional aggregates with an architecture and functional activity similar to native islets: pseudoislets. Pseudoislets represent an alternative source for islet transplantation, because their transplant results in a long term allograft acceptance
Derek J McKillop et al.
Journal of agricultural and food chemistry, 51(15), 4382-4388 (2003-07-10)
To provide a tool to study folate bioavailability under controlled conditions, a methodology was developed to produce extracts representative of natural food folates but removed from their matrix and sufficiently concentrated so as to elicit a response in biomarkers of
Jonathan R Weinstein et al.
Glia, 56(1), 16-26 (2007-10-03)
Lipopolysaccharide (LPS/endotoxin) is a potent immunologic stimulant. Many commercial-grade reagents used in research are not screened for LPS contamination. LPS induces a wide spectrum of proinflammatory responses in microglia, the immune cells of the brain. Recent studies have demonstrated that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service