Skip to Content
Merck
All Photos(7)

Documents

S7571

Sigma-Aldrich

Superoxide Dismutase from bovine erythrocytes

greener alternative

lyophilized powder, ≥3,000 units/mg protein, Protein ≥95 % by biuret

Synonym(s):

CU/ZN-SOD, Superoxide Dismutase 1 bovine, cytocuprein, erythrocuprein, hemocuprein, SOD, Superoxide: superoxide oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine

form

lyophilized powder

specific activity

≥3,000 units/mg protein

mol wt

32.5 kDa

composition

Protein, ≥95% biuret

storage condition

(Store under nitrogen.
Tightly closed. Dry.)

greener alternative product characteristics

Atom Economy
Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

immunoblotting: suitable
inhibition assay: suitable

color

blue-green

pI 

4.95

solubility

water: 20 mg/mL
aqueous buffer, pH 7.5: soluble

UniProt accession no.

application(s)

diagnostic assay manufacturing

greener alternative category

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling

Superoxide dismutase (SOD) is a redox-active metalloenzyme expressed in both aerobic and anaerobic living organisms. Bovine superoxide dismutase or CuZn SOD is a homodimer with each subunit containing one zinc and one copper ion.

Application

Superoxide dismutase from bovine erythrocytes has been used:

  • in a study to assess a kinetic model of radiation-induced inactivation of superoxide dismutase in nitrous oxide-saturated solutions
  • in a study to investigate the possible participation of superoxide anion in the intestinal tryptophan 2,3-dioxygenase reaction
  • to investigate its effect on the hemolysis rate of human RBCs and hemoglobin-nitric oxide complex (HbNO) stability in human erythrocytes
  • in combination with catalase to study its effect on cell differentiation in vitro
  • to quantify superoxide levels and study their effect on reactivity in mouse pulmonary arteries through chemiluminescence and cytochrome C reduction methods

Biochem/physiol Actions

Superoxide Dismutase from bovine erythrocytes catalyzes the dismutation of superoxide radicals to hydrogen peroxide and molecular oxygen. It serves as an antioxidant and plays a critical role in the defense of cells against the toxic effects of oxygen radicals. Competes with nitric oxide (NO) for superoxide anion (which reacts with NO to form peroxynitrite), thereby SOD promotes the activity of NO. SOD has also been shown to suppress apoptosis in cultured rat ovarian follicles, neural cell lines, and transgenic mice.

Unit Definition

One unit will inhibit reduction of cytochrome c by 50% in a coupled system with xanthine oxidase at pH 7.8 at 25 °C in a 3.0 ml reaction volume. Xanthine oxidase concentration should produce an initial ΔA550 of 0.025 ± 0.005 per min.

Physical form

Lyophilized powder, essentially salt-free

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Possible participation of superoxide anion in the intestinal tryptophan 2,3-dioxygenase reaction.
F Hirata et al.
The Journal of biological chemistry, 246(24), 7825-7826 (1971-12-25)
Michael A Hough et al.
Structure (London, England : 1993), 11(8), 937-946 (2003-08-09)
Copper zinc superoxide dismutase (CuZnSOD) forms a crucial component of the cellular response to oxidative stress by catalyzing the dismutation of the superoxide radical to hydrogen peroxide and water. Mutations in human CuZnSOD are associated with the development of familial
Sebastian Overmans et al.
Aquatic toxicology (Amsterdam, Netherlands), 198, 165-174 (2018-03-20)
Exposure to polycyclic aromatic carbons (PAHs) poses a growing risk to coral reefs due to increasing shipping and petroleum extraction in tropical waters. Damaging effects of specific PAHs can be further enhanced by the presence of ultraviolet radiation, known as
Colleen M Hansel et al.
mBio, 10(2) (2019-03-14)
There is a growing appreciation within animal and plant physiology that the reactive oxygen species (ROS) superoxide is not only detrimental but also essential for life. Yet, despite widespread production of extracellular superoxide by healthy bacteria and phytoplankton, this molecule
Superoxide dismutases and superoxide reductases.
Yuewei Sheng et al.
Chemical reviews, 114(7), 3854-3918 (2014-04-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service