Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Superoxide Dismutase from bovine erythrocytes
Synonym(s):
SOD, Superoxide: superoxide oxidoreductase
CAS Number:
9054-89-1
EC Number:
232-943-0
Products
Brand
Sigma-Aldrich (3)
Sustainability
Biological Source
Color
Feature
Physical Form
pK Value
Product Line
Purity
Tissue
Chemical Composition
Application
Relevant Disease
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S7571
Superoxide Dismutase from bovine erythrocytes,
lyophilized powder, ≥3,000 units/mg protein, Protein ≥95 % by biuret
Expand
View Pricing
S5395
Superoxide Dismutase from bovine erythrocytes,
BioReagent, ≥3,000 units/mg protein, lyophilized powder, suitable for cell culture
Expand
View Pricing
S7446
Superoxide Dismutase from bovine erythrocytes,
BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Expand
View Pricing