S6765
Schenk and Hildebrandt Basal Salt Mixture
fine powder, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.72
Recommended Products
form
fine powder
Quality Level
technique(s)
cell culture | plant: suitable
application(s)
agriculture
storage temp.
2-8°C
Application
Schenk and Hildebrandt Vitamin 100 × (S3766) and Schenk and Hildebrandt Basal Salt Mixture (S6765) are chemstocks that may be used to generate Schenk and Hildebrandt (SH) medium for use in plant cell culture and microproagation.
Formula variant
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Sol. 3
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sung Jin Hwang
Methods in molecular biology (Clifton, N.J.), 547, 263-273 (2009-06-13)
Hairy root clones of Rehmannia glutinosa were established via transformation with Agrobacterium rhizogenes ATCC15834. To optimize the culturing conditions for both root growth and catalpol production, the effects of various combinations of seven basal media, pH, and carbon sources were
Monya M Costa et al.
Biotechnology letters, 30(7), 1265-1270 (2008-02-15)
Six-year-old Levisticum officinale (lovage) hairy root cultures were used to study the effect of eight different NH(4) (+):NO(3) (-) ratios on their growth and volatile components. All cultures were kept at 24 degrees C on orbital shakers at 80 rpm
Suk Weon Kim et al.
Methods in molecular biology (Clifton, N.J.), 547, 107-115 (2009-06-13)
This chapter describes culture conditions for high-frequency plant regeneration via somatic embryogenesis and cryopreservation from cell suspension cultures of Ranunculus kazusensis. Zygotic embryos form white nodular structures and pale-yellow calli at a frequency of 84.9% on half-strength Schenk and Hildebrandt
Inês S Nunes et al.
Planta medica, 75(4), 387-391 (2009-01-22)
The biotransformation capacity of Levisticum officinale W.D.J. Koch hairy root cultures was studied by evaluating the effect of the addition of 25 mg/L menthol or geraniol on morphology, growth, and volatiles production. L. officinale hairy root cultures were maintained for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service