Skip to Content
Merck
All Photos(1)

Key Documents

S6760

Sigma-Aldrich

Span® 80

nonionic surfactant

Synonym(s):

Sorbitane monooleate, sorbitan oleate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

mol wt

428.62 g/mol

concentration

≥60% (GC)

HLB

4.3

application(s)

microbiology

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@@H](O)[C@H]1OC[C@H](O)[C@H]1O

InChI

1S/C24H44O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(27)29-19-21(26)24-23(28)20(25)18-30-24/h9-10,20-21,23-26,28H,2-8,11-19H2,1H3/b10-9-/t20-,21+,23+,24+/m0/s1

InChI key

NWGKJDSIEKMTRX-AAZCQSIUSA-N

General description

Span® 80 is a nonionic surfactant.

Application

Span® 80 has been used in a study to assess transfersomes as a transdermal delivery system for sertraline. It has also been used in a study to investigate the dominant factors affecting the stability of nanoemulsions through the use of artificial neural networks (ANNs).

Other Notes

Fatty acid composition: Oleic acid (C18:1) ≤ 60%; balance primarily linoleic (C18:2), linolenic (C18:3) and palmitic (C16:0) acids.

Legal Information

Span is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ankit Gupta et al.
Scientia pharmaceutica, 80(4), 1061-1080 (2012-12-25)
The aim of the present study was to investigate transfersomes as a transdermal delivery system for the poorly soluble drug, sertraline, in order to overcome the troubles associated with its oral delivery. Different transfersomal formulations were prepared with non-ionic surfactant
Jingchun Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 28(17), 6769-6775 (2012-04-06)
The effects of salt on emulsions containing sorbitan oleate (Span 80) and Laponite particles were investigated. Surprisingly, a novel double phase inversion was induced by simply changing the salt concentration. At fixed concentration of Laponite particles in the aqueous phase
Keita Hayashi et al.
International journal of biological sciences, 9(2), 142-148 (2013-02-16)
Span 80 (sorbitan monooleate) vesicles behaved differently from conventional phospholipid vesicles (liposomes) because the former had a more fluid interface. After doxorubicin hydrochloride (DOX) was encapsulated into the Span 80 vesicle (loading efficiency: 63 %), DOX-loaded Span 80 vesicles (DVs)
Jean-François Cotte et al.
International journal of pharmaceutics, 436(1-2), 233-239 (2012-06-21)
Mass spectrometry (MS) and high performance liquid chromatography coupled to mass spectrometry (HPLC-MS) techniques were developed to characterize two surfactants, cetheareth-12 and sorbitan oleate, used to manufacture AF03, an emulsified oil-in-water (O/W) adjuvant. MS was first used to characterize the
Hoang Hirschberg et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 46(1-2), 1-7 (2012-02-15)
In the search for an optimal approach for the transcutaneous immunization (TCI) of hepatitis B surface antigen (HBsAg), two vesicle formulations, L595 vesicles (composed of sucrose-laurate ester and octaoxyethylene-laurate ester) and sPC vesicles (composed of soybean-phosphatidylcholine and Span-80) were prepared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service