Skip to Content
Merck
All Photos(2)

Key Documents

S3386

Sigma-Aldrich

Sorbitan sesquioleate

nonionic surfactant

Synonym(s):

Arlacel® 83, Span® 83

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

corn

Quality Level

vapor pressure

0.81 psi ( 20 °C)

form

liquid

refractive index

n20/D 1.478 (lit.)

density

0.989 g/mL at 25 °C (lit.)

functional group

ester

lipid type

oils

shipped in

ambient

storage temp.

room temp

SMILES string

OC[C@@H](O)C1OC[C@H](O)[C@H]1O.OC[C@@H](O)C2OC[C@H](O)[C@H]2O.CCCCCCCC\C=C/CCCCCCCC(O)=O.CCCCCCCC\C=C/CCCCCCCC(O)=O.CCCCCCCC\C=C/CCCCCCCC(O)=O

InChI

1S/3C18H34O2.2C6H12O5/c3*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;2*7-1-3(8)6-5(10)4(9)2-11-6/h3*9-10H,2-8,11-17H2,1H3,(H,19,20);2*3-10H,1-2H2/b3*10-9-;;/t;;;2*3-,4+,5-,6-/m...11/s1

InChI key

SPFRYHCSFBFNTR-RKGWDQTMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sorbitan sesquioleate (SSO) is a sorbitol derivative emulsifier. It is a common emulsifier component in topical corticosteroids, topical antibiotics, topical antifungals, moisturizing creams and lotions and topical retinoids.

Biochem/physiol Actions

Sorbitan sesquioleate is useful as a stabilizing agent in polymerization process to produce spherical acrylamide beads. It has been used in many topical agents. SSO have recently been implicated in allergic dermatitis.

Other Notes

Fatty acid composition: Oleic acid (C18:1) approx. 70%; balance primarily palmitic acid (C16:0), stearic acid (C18:0) and linoleic acid (C18:2).

Legal Information

Arlacel is a registered trademark of Croda International PLC
Span is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sorbitan sesquioleate.
Mari Paz Castanedo-Tardan et al.
Dermatitis : contact, atopic, occupational, drug, 19(4), E22-E23 (2008-08-05)
Contact urticaria syndrome from sorbitan sesquioleate in a corticosteroid ointment.
M Hardy et al.
Contact dermatitis, 32(6), 360-361 (1995-06-01)
S Fukanoki et al.
Avian diseases, 44(4), 869-873 (2001-02-24)
The relationship between the antigen release from formulations in vitro and the antibody response after administration of water-in-oil-in-water (W/O/W) emulsion vaccines containing Haemophilus paragallinarum (Hpg) was studied in chickens. Increases of sorbitan sesquioleate volume in the formulation led to slower
Sorbitan sesquioleate as an allergen.
D I Orton et al.
Contact dermatitis, 44(3), 190-191 (2001-02-24)
The Validity of Patch Testing
Lachapelle JM, et al.
Patch Testing Tips, 49(4), 37-61 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service