Skip to Content
Merck
All Photos(1)

Key Documents

S5921

Sigma-Aldrich

Staurosporine

from Streptomyces sp., ≥95% (HPLC), film, protein kinase inhibitor

Synonym(s):

protein kinase c inhibitor, staurosporine, Antibiotic AM-2282

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H26N4O3
CAS Number:
Molecular Weight:
466.53
Beilstein:
1060573
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Product Name

Staurosporine from Streptomyces sp., for molecular biology, ≥95% (HPLC)

grade

Molecular Biology
for molecular biology

Quality Level

Assay

≥95% (HPLC)

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

CN[C@@H]1C[C@@H]2O[C@@](C)([C@@H]1OC)n3c4ccccc4c5c6CNC(=O)c6c7c8ccccc8n2c7c35

InChI

1S/C28H26N4O3/c1-28-26(34-3)17(29-2)12-20(35-28)31-18-10-6-4-8-14(18)22-23-16(13-30-27(23)33)21-15-9-5-7-11-19(15)32(28)25(21)24(22)31/h4-11,17,20,26,29H,12-13H2,1-3H3,(H,30,33)/t17-,20?,26-,28+/m1/s1

InChI key

HKSZLNNOFSGOKW-ZGQXJOJZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: indol derivative
Staurosporine is an alkaloid that is obtained from Streptomyces sp. It possesses a wide range of activities such as protein kinase C inhibitor, inhibition of tumor cell proliferation, and blockage of cell cycle progression in various cells. Staurosporine is a well-known inducer of apoptosis in various cell lines. The mechanisms of staurosporine-induced apoptosis vary among different cell lines that include, mitochondrial apoptotic pathway, Bcl-2 overexpression, and caspase-independent mechanisms. It partially reverses multi-drug resistance (MDR), sensitizing cells with MDR phenotype to cytotoxic agents. Staurosporine inhibits p-glycoprotein (Pgp) phosphorylation. However, the functional significance of Pgp phosphorylation is ill-defined.

Application

Staurosporine has been used as an apoptosis inducer in apoptosis assay in human colorectal cancer cell lines and to evaluate the caspase 3 and 7 activity in endothelial cell line. It has also been used as a positive control in apoptosis assay in human umbilical vein endothelial cells.

Biochem/physiol Actions

Potent inhibitor of phospholipid/calcium-dependent protein kinase. Inhibits the upregulation of VEGF expression in tumor cells.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Analysis Note

Tested for inhibition of interleukin 2 production in Jurkat cells.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 1B - Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Forsthuber et al.
Environmental pollution (Barking, Essex : 1987), 293, 118543-118543 (2021-11-21)
Perfluorooctane sulfonic acid (PFOS) is a ubiquitous environmental pollutant. In humans, PFOS exposure has been associated with a number of adverse health outcomes, including reduced birth weight. Whether PFOS is capable of affecting angiogenesis and thus possibly fetal development is
H J Chae et al.
Pharmacological research, 42(4), 373-381 (2000-09-16)
Staurosporine, a microbial alkaloid, is a strong inhibitor of protein kinases. We induced apoptosis in murine osteoblast MC3T3E-1 cells by exposure to the staurosporine. Staurosporine transiently increased the phosphotransferase activity of c-Jun N-terminal kinase-1 (JNK1), which in turn may activate
Xu Dong Zhang et al.
Molecular cancer therapeutics, 3(2), 187-197 (2004-02-27)
Staurosporine has long been used in vitro as an initiator of apoptosis in many different cell types, but the mechanism involved remains poorly understood. In the present study, we have examined the apoptosis-inducing potential of staurosporine in cultured melanoma cell
A Gescher
General pharmacology, 31(5), 721-728 (1998-11-11)
1. Protein kinase C (PKC) is a family of serine/threonine-directed protein kinases that are pivotal regulators of cellular growth, transformation and death. PKC has therefore been considered to be a suitable target for novel antineoplastic drugs. 2. Twenty years ago
S Ståhl et al.
Cell death & disease, 4, e454-e454 (2013-01-11)
Radiation therapy is frequently used to treat non-small cell lung cancers (NSCLCs). We have previously shown that a combination of ionizing radiation (IR) and the staurosporine analog PKC 412, but not Ro 31-8220, increases cell death in NSCLC cells. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service