Skip to Content
Merck
All Photos(2)

Documents

37095

Supelco

Staurosporine

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H26N4O3
CAS Number:
Molecular Weight:
466.53
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CN[C@@H]1C[C@@H]2O[C@@](C)([C@@H]1OC)n3c4ccccc4c5c6CNC(=O)c6c7c8ccccc8n2c7c35

InChI

1S/C28H26N4O3/c1-28-26(34-3)17(29-2)12-20(35-28)31-18-10-6-4-8-14(18)22-23-16(13-30-27(23)33)21-15-9-5-7-11-19(15)32(28)25(21)24(22)31/h4-11,17,20,26,29H,12-13H2,1-3H3,(H,30,33)/t17-,20?,26-,28+/m1/s1

InChI key

HKSZLNNOFSGOKW-ZGQXJOJZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Staurosporine belongs to the class of indolocarbazole alkaloids and shows anti-fungal and anti-oomycete activities. It is isolated from Streptomyces sp. and is known to be a broad-spectrum inhibitor of protein kinase.

Application


  • Protein Painting for Structural Analysis: This research utilizes staurosporine′s role in protein kinase inhibition to develop a technique called protein painting, which is essential for structural and binding site analysis. This approach enhances understanding of protein interactions and can be pivotal in drug design, particularly in targeting specific kinase enzymes in cancer research (Zheng Z et al., 2024).

  • Identification of Anthelmintic Activities: Staurosporine is employed to identify small molecule inhibitors that target conserved proteins among gastrointestinal nematodes. This study not only highlights its role in parasitic infection treatments but also its broader applications in developing antiparasitic drugs, leveraging its properties as a potent kinase inhibitor (Jung H et al., 2024).

  • Development of Novel Cytoprotective Compounds: Focusing on mitochondria-dependent cell death, this research explores staurosporines potential to inhibit pathways involved in cellular apoptosis. This is particularly crucial in developing therapeutic strategies against diseases characterized by unwanted cell death, such as neurodegenerative diseases and certain types of cancer (Matsuyama M et al., 2023).

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Anestål et al.
PloS one, 3(4), e1846-e1846 (2008-04-03)
SecTRAPs (selenium compromised thioredoxin reductase-derived apoptotic proteins) can be formed from the selenoprotein thioredoxin reductase (TrxR) by targeting of its selenocysteine (Sec) residue with electrophiles, or by its removal through C-terminal truncation. SecTRAPs are devoid of thioredoxin reductase activity but
A Gescher
General pharmacology, 31(5), 721-728 (1998-11-11)
1. Protein kinase C (PKC) is a family of serine/threonine-directed protein kinases that are pivotal regulators of cellular growth, transformation and death. PKC has therefore been considered to be a suitable target for novel antineoplastic drugs. 2. Twenty years ago
S Ståhl et al.
Cell death & disease, 4, e454-e454 (2013-01-11)
Radiation therapy is frequently used to treat non-small cell lung cancers (NSCLCs). We have previously shown that a combination of ionizing radiation (IR) and the staurosporine analog PKC 412, but not Ro 31-8220, increases cell death in NSCLC cells. To
Staurosporine, K-252 and UCN-01: potent but nonspecific inhibitors of protein kinases.
U T Rüegg et al.
Trends in pharmacological sciences, 10(6), 218-220 (1989-06-01)
Osman A B S M Gani et al.
Natural product reports, 27(4), 489-498 (2010-03-26)
The isolation in 1977 of the microbial alkaloid staurosporine inaugurated research into several distinct series of related natural and synthetic compounds. This has especially included research into applications as anticancer drugs, beginning with the observation of low nanomolar inhibition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service