Skip to Content
Merck
All Photos(2)

Documents

S4322

Sigma-Aldrich

Anti-Sirt6 (N-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NAD-dependent deacetylase sirtuin 6, Anti-SIR2-like protein 6, Anti-SIR2L6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

rat (predicted), mouse, human

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

western blot: 2-4 μg/mL using whole extracts of human 293 cells transfected with Sirt6
western blot: 2-4 μg/mL using whole extracts of mouse 3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIRT6(51548)
rat ... Sirt6(299638)

General description

Sirtuin 6 (Sirt6) is a nuclear and chromatin-associated protein that belongs to the mammalian Sir2 gene family. It is broadly expressed with the highest levels in muscle, brain and heart.

Specificity

Anti-Sirt6 (N-terminal) recognizes human and mouse Sirt6. The detection of the Sirt6 band by immunoblotting is specifically inhibited with the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 19-33 of human Sirt6 via a C-terminal cysteine residue. The sequence is identical in mouse and differs by one amino acid in rat.

Application

Anti-Sirt6 (N-terminal) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Sirtuin 6 (Sirt6) plays a key role in DNA repair and maintenance of genomic stability in cells. Sirt6 is enzymatically inactive as a NAD+ -dependent protein deacetylase but displays auto-ADP-ribosyltransferase activity. Loss of Sirt6 leads to the development of an acute degenerative aging-like phenotype.

Physical form

Solution in 0.01 M phosphate buffered, saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential expression of sirtuin family members in the developing, adult, and aged rat brain
Sidorova-Darmos E, et al.
Frontiers in Aging Neuroscience, 6 (2014)
Sergii Domanskyi et al.
Apoptosis : an international journal on programmed cell death, 22(11), 1336-1343 (2017-09-01)
Apoptosis is essential for numerous processes, such as development, resistance to infections, and suppression of tumorigenesis. Here, we investigate the influence of the nutrient sensing and longevity-assuring enzyme SIRT6 on the dynamics of apoptosis triggered by serum starvation. Specifically, we
Mouse Sir2 Homolog SIRT6 Is a Nuclear ADP-ribosyltransferase
Liszt G, et al.
Test, 280 (2005)
Genomic Instability and Aging-like Phenotype in the Absence of Mammalian SIRT6
Mostoslavsky R, et al.
Cell, 124 (2006)
Justin W Nicholatos et al.
Acta neuropathologica communications, 6(1), 120-120 (2018-11-10)
Parkinson's disease is characterized by progressive death of dopaminergic neurons, leading to motor and cognitive dysfunction. Epidemiological studies consistently show that the use of tobacco reduces the risk of Parkinson's. We report that nicotine reduces the abundance of SIRT6 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service