Skip to Content
Merck
All Photos(1)

Key Documents

R0503

Sigma-Aldrich

Reactive Red 120−Agarose

saline suspension, Type 3000-CL

Synonym(s):

Reactive Red Agarose, Red 120-Agarose, Red Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

biological source

plant

Quality Level

type

Type 3000-CL

form

saline suspension

extent of labeling

≥3 μmol per mL

technique(s)

affinity chromatography: suitable

matrix

cross-linked 4% beaded agarose

suitability

suitable for chromatography

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Reactive Red 120-agarose is used in affinity chromatography, protein chromatography and dye resins. Reactive Red 120-agarose has been used to study wheat quality breeding as well as to provide strong evidence that purified human P-glycoprotein (Pgp) functions as an ATP-dependent drug transporter.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

P De Prada et al.
Applied and environmental microbiology, 63(7), 2928-2931 (1997-07-01)
Two extracellular, heat-labile alkaline phosphatases were purified from a psychrophilic Arthrobacter isolate, D10. The enzymes were active over different pH ranges, used distinct substrates, and had different kinetic properties. Each enzyme reacted specifically to its own antibody during immunoblot analysis.
Koji Iwamoto et al.
Plant physiology, 133(2), 893-900 (2003-09-16)
Mannitol-1-phosphate (M1P) dehydrogenase (M1PDH; EC 1.1.1.17), an enzyme catalyzing the reduction of Fru-6-phosphate (F6P) to M1P in algal mannitol biosynthesis, was purified to homogeneity from a cell homogenate of the eulittoral red alga Caloglossa continua (Okamura) King et Puttock. The
Removing trace fluorescent contaminants from GroEL preparations.
F Weber
Methods in molecular biology (Clifton, N.J.), 140, 63-64 (2001-08-04)
Gregory A Bannikov et al.
American journal of veterinary research, 68(9), 995-1004 (2007-09-04)
To characterize and purify covalent complexes of matrix metalloproteinase-9 (MMP-9) and haptoglobin released by bovine granulocytes in vitro. Blood samples obtained from healthy cows and cows with acute and chronic inflammation to obtain WBCs and sera. WBCs were isolated by
P C Larosa et al.
Plant physiology, 96(1), 245-250 (1991-05-01)
Tobacco (Nicotiana tabacum L. var Wisconsin 38) cells that are adapted to 428 millimolar NaCl accumulate proline mainly due to increased synthesis from glutamate. These cells were used to evaluate the possible role of Delta(1)-pyrroline-5-carboxylate reductase in the regulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service