Skip to Content
Merck
All Photos(1)

Key Documents

S4047

Sigma-Aldrich

Anti-S6 Kinase antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-p70S6k

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen 70 kDa

contains

15 mM sodium azide

species reactivity

rat, mouse, human

technique(s)

microarray: suitable
western blot: 1:10,000 using mouse NIH 3T3 fibroblast cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

S6 kinases are Ser/Thr protein kinases that are capable of phosphorylating the 40S ribosomal protein S6 in response to mitogen stimulation. S6 kinase (p70s6k) is activated by an upstream kinase, mTOR in response to nutrients and growth factors. S6 kinase (p70s6k) is a critical regulatory component and mTOR along with the eukaryotic initiation factor 4E (eIF4E)–binding protein 1 (4E-BP1) and S6 kinase (p70s6k) leads to increased ribosomal biogenesis and hence translation. As a critical component and has the ability to modify the cellular responses to availability of nutrients, and growth factors like insulin and IGF-1 when mTOR is chronically active. In presence of excess and chronic insulin, or high fat diets, S6 kinase (p70s6k) regulates by a negative feedback loop and constitutively shuts down the responsiveness of cells to insulin. Thus mTOR through the S6 kinase (p70s6k) plays a pivotal role in the development of insulin resistance and type 2 diabetes. mTOR signaling via p70S6 kinase also drives cell growth and proliferation, cancer and aging.
Anti-S6 Kinase reacts specifically with S6 kinase (70 kDa) in human, rat and mouse.

Specificity

The sequence of the immunizing peptide is identical in rat and mouse S6K.

Immunogen

synthetic peptide corresponding to amino acids 6-23 of human S6 kinase.

Application

Anti- S6 kinase antibody may be used for detection by immunoblotting at a working dilution of 1:10,000 using mouse NIH 3T3 fibroblast cell extract. Detection by immunoblotting was also done in primary cultures of mouse glial cells and in LnCAP, prostate cancer cell line.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Zoncu et al.
Nature reviews. Molecular cell biology, 12(1), 21-35 (2010-12-16)
In all eukaryotes, the target of rapamycin (TOR) signalling pathway couples energy and nutrient abundance to the execution of cell growth and division, owing to the ability of TOR protein kinase to simultaneously sense energy, nutrients and stress and, in
Jianying Guo et al.
Oncology letters, 22(2), 630-630 (2021-07-17)
Colorectal cancer (CRC) is the fourth most lethal cancer in the world. Heat shock protein 60 (HSP60), a mitochondrial chaperone that maintains mitochondrial proteostasis, is highly expressed in tumors compared with in paracancerous tissues, suggesting that high HSP60 expression benefits
W L See et al.
Oncogene, 29(12), 1720-1731 (2010-01-12)
The tumor suppressive activities of the Kip-family of cyclin-dependent kinase (cdk) inhibitors often go beyond their role directly regulating the cell cycle. In this study, we show that p27 enhances Rad51 accumulation during repair of double-strand DNA breaks. Progression of
K Hara et al.
The Journal of biological chemistry, 273(23), 14484-14494 (1998-06-11)
The present study identifies the operation of a signal tranduction pathway in mammalian cells that provides a checkpoint control, linking amino acid sufficiency to the control of peptide chain initiation. Withdrawal of amino acids from the nutrient medium of CHO-IR
Hongyu Zhou et al.
Current protein & peptide science, 11(6), 409-424 (2010-05-25)
The mammalian target of rapamycin (mTOR) has attracted substantial attention because of its involvement in a variety of diseases, such as cancer, cardiac hypertrophy, diabetes and obesity. Current knowledge indicates that mTOR functions as two distinct multiprotein complexes, mTORC1 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service