S3382
Solvent Blue 38
Practical grade, powder
Synonym(s):
Luxol® Fast Blue MBSN
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Product Name
Solvent Blue 38, practical grade
grade
practical grade
form
powder
technique(s)
titration: suitable
solubility
methanol: 1 mg/mL, blue
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Solvent Blue 38 is a phthalocyanine stain, used for staining myelin, neurons and glial fibres.
Application
Solvent Blue 38 has been used to stain tissue sections to assess remyelination.
Biochem/physiol Actions
Solvent Blue 38 exhibits a strong affinity towards bases in the lipoproteins of myelin sheath due to which it is stained blue.
Legal Information
Luxol is a registered trademark of Rohm and Haas Co.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Kazuya Kitamura et al.
PloS one, 6(11), e27706-e27706 (2011-12-06)
Many therapeutic interventions for spinal cord injury (SCI) using neurotrophic factors have focused on reducing the area damaged by secondary, post-injury degeneration, to promote functional recovery. Hepatocyte growth factor (HGF), which is a potent mitogen for mature hepatocytes and a
Stereotactic Administration of Edaravone Ameliorates Collagenase-Induced Intracerebral Hemorrhage in Rat
Yan Zhang
CNS Drugs (2016)
Cornelia Laule et al.
NeuroImage, 40(4), 1575-1580 (2008-03-07)
Myelin water imaging (MWI) promises to be invaluable in understanding neurological diseases like MS. However, a limitation of MWI is signal to noise ratio. Recently, a number of investigators have performed MWI at field strengths higher than 1.5 T. Our
M Lindner et al.
Neuropathology and applied neurobiology, 34(1), 105-114 (2007-10-27)
To understand the mechanisms of remyelination and the reasons for regeneration failure is one of the major challenges in multiple sclerosis research. This requires a good knowledge and reliable analysis of experimental models. This work was undertaken to characterize the
Collagen scaffolds modified with CNTF and bFGF promote facial nerve regeneration in minipigs
Yi Cui
Biomaterials research (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service