Skip to Content
Merck
All Photos(1)

Key Documents

S3065

Sigma-Aldrich

SC 19220

≥98% (HPLC), solid

Synonym(s):

2-Acetylhydrazide 10(11H)-carboxylic acid, 8-Chloro-dibenz[b,f][1,4]oxazepine-10(11H)-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N3O3Cl
CAS Number:
Molecular Weight:
331.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

mp

190-191 °C (lit.)

solubility

DMSO: soluble 10 mg/mL
H2O: insoluble

SMILES string

CC(=O)NNC(=O)N1Cc2ccccc2Oc3ccc(Cl)cc13

InChI

1S/C16H14ClN3O3/c1-10(21)18-19-16(22)20-9-11-4-2-3-5-14(11)23-15-7-6-12(17)8-13(15)20/h2-8H,9H2,1H3,(H,18,21)(H,19,22)

InChI key

KNURFLJTOUGOOQ-UHFFFAOYSA-N

Gene Information

human ... PTGER1(5731)
rat ... Ptger1(25637)

Application

SC 19220 was used to study the role of prostaglandin E2 in oxygen-glucose deprivation-induced neurotoxicity and preconditioning-induced neuroprotection in rat cortical cultures.

Biochem/physiol Actions

SC 19220 is a competitive antagonist of prostaglandin E receptors. It competes with PGE2 in regulating vesicourethral motility and increases the bladder capacity of rats. It inhibits the contraction of smooth muscles in response to prostaglandins E2 and F. SC 19220 also inhibits the formation of osteoclasts induced by vitamin D3, parathyroid hormone, IL-11 and IL-6.
EP1 Prostanoid receptor antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Inoue et al.
The Journal of endocrinology, 161(2), 231-236 (1999-05-13)
1,25 Dihydroxy vitamin D3 (1,25(OH)2D3), prostaglandin (PG) E2 and parathyroid hormone (PTH) induce osteoclast formation in cell cultures. Previously, we have shown that SC-19220, an antagonist of the EP1 subtype of PGE receptors, inhibited tartrate-resistant acid phosphatase (TRAP)-positive cell formation
Alexander V Timoshenko et al.
Experimental cell research, 289(2), 265-274 (2003-09-23)
Aberrant upregulation of COX-2 enzyme resulting in accumulation of PGE2 in a cancer cell environment is a marker for progression of many cancers, including breast cancer. Four subtypes of cell surface receptors (EP1, EP2, EP3, and EP4), which are coupled
C A Maggi et al.
European journal of pharmacology, 152(3), 273-279 (1988-08-02)
SC-19220 (5-20 mg/kg i.v.), a competitive receptor antagonist of PGE, increased the bladder capacity and reduced the voiding efficiency of micturition (elicited by slow transvesical filling) of urethane-anesthetized rats. The effect of SC-19220 was prevented by indomethacin pretreatment, whereas indomethacin
Xavier Norel et al.
British journal of pharmacology, 142(4), 788-796 (2004-06-03)
1. In human pulmonary vascular preparations, precontracted arteries were more sensitive to the relaxant effect of acetylcholine (ACh) than veins (pD(2) values: 7.25+/-0.08 (n=23) and 5.92+/-0.09 (n=25), respectively). Therefore, the role of prostacyclin (PGI(2)) was explored to examine whether this
Micaela Zonta et al.
The Journal of physiology, 553(Pt 2), 407-414 (2003-09-23)
The synaptic release of glutamate evokes in astrocytes periodic increases in [Ca2+]i, due to the activation of metabotropic glutamate receptors (mGluRs). The frequency of these [Ca2+]i oscillations is controlled by the level of neuronal activity, indicating that they represent a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service