Skip to Content
Merck
All Photos(1)

Key Documents

S1439

Sigma-Aldrich

Monoclonal Anti-SERCa2 ATPase antibody produced in mouse

clone IID8

Synonym(s):

Monoclonal Anti-Sarcoplasmic or Endoplasmic Reticulum Ca2+ ATPase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

antibody form

purified from hybridoma cell culture

Quality Level

antibody product type

primary antibodies

clone

IID8, monoclonal

species reactivity

bovine, canine, rat, human, pig, rabbit

technique(s)

immunocytochemistry: suitable
immunohistochemistry (frozen sections): 1:500
indirect immunofluorescence: suitable
western blot: 1:500-1:5,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP2A2(488)
rat ... Atp2a2(29693)

Immunogen

purified canine cardiac sarcoplasmic reticulum.

Physical form

Supplied as a solution in PBS containing 0.05% sodium azide.

Other Notes

Not suitable for immunoblotting in rat SERCA2.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Li et al.
Traffic (Copenhagen, Denmark), 18(4), 232-241 (2017-02-06)
Darier's disease (DD) is an autosomal dominantly inherited skin disorder caused by mutations in sarco/endoplasmic reticulum Ca2+ -ATPase 2 (SERCA2), a Ca2+ pump that transports Ca2+ from the cytosol to the endoplasmic reticulum (ER). Loss of desmosomes and keratinocyte cohesion
Victor Galvan-Alvarez et al.
Redox biology, 66, 102859-102859 (2023-09-05)
When high-intensity exercise is performed until exhaustion a "functional reserve" (FR) or capacity to produce power at the same level or higher than reached at exhaustion exists at task failure, which could be related to reactive oxygen and nitrogen species
Haibin Gong et al.
Experimental and therapeutic medicine, 12(3), 1355-1362 (2016-09-08)
The study was conducted to examine the effects of ICI 118,551 on the systolic function of cardiac muscle cells of rats in heart failure and determine the molecular mechanism of selective β2-adrenergic receptor (β2-AR) antagonist on these cells. The chronic
Luca Hegedũs et al.
International journal of cancer, 140(12), 2758-2770 (2016-11-05)
Oncogenic mutations of BRAF lead to constitutive ERK activity that supports melanoma cell growth and survival. While Ca2+ signaling is a well-known regulator of tumor progression, the crosstalk between Ca2+ signaling and the Ras-BRAF-MEK-ERK pathway is much less explored. Here
Luca Hegedüs et al.
Frontiers in oncology, 7, 95-95 (2017-06-10)
Several new therapeutic options emerged recently to treat metastatic melanoma; however, the high frequency of intrinsic and acquired resistance among patients shows a need for new therapeutic options. Previously, we identified the plasma membrane Ca2+ ATPase 4b (PMCA4b) as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service