Skip to Content
Merck

TRYPSEQM-RO

Roche

Trypsin Sequencing Grade, modified

from bovine pancreas

Synonym(s):

Trypsin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Biological source:
bovine pancreas
Concentration:
0.01-0.2 % (w/w)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine pancreas

form

lyophilized (salt-free)

mol wt

24.000 g/mol

packaging

pkg of 4 × 100 μg (11418033001), pkg of 4 × 25 μg (11418025001)

manufacturer/tradename

Roche

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

concentration

0.01-0.2 % (w/w)

technique(s)

protein sequencing: suitable

impurities

Chymotrypsin

color

white

optimum pH

8.0

solubility

10 g/L

suitability

suitable for protein modification

UniProt accession no.

application(s)

life science and biopharma

foreign activity

Contaminating activities corresponds
Chymotrypsin , contains

storage temp.

2-8°C

Gene Information

cow ... PRSS1(780933)

General description

Trypsin Sequencing Grade, modified, is isolated from bovine pancreas as a highly purified and specific protease, and subsequently modified.

Trypsin is a highly efficient and specific protease widely used in proteomics for protein digestion. It produces short peptides with specific characteristics that are compatible with current separation and identification methods such as liquid chromatography, mass spectrometry (MS).

Inhibitors:
TLCK, DFP, PMSF, leupeptin, soybean trypsin inhibitor, trypsin inhibitor from hen egg, aprotinin, α2-macroglobulin,α1-antitrypsin, APMSF, and antipain.

Application

Use Trypsin Sequencing Grade, modified, to generate glycopeptides from purified glycoproteins.
It is used for:
  • Protein-structure elucidation
  • Tryptic mapping
  • Fingerprinting analysis
  • Sequence analysis
  • Translocation studies
  • Protein identification
  • Protein digestion during lipoprotein preparation for liquid chromatography-tandem mass spectrometry (LC-MS/MS)

Biochem/physiol Actions

Trypsin is a serine endopeptidase. At pH 7.5–9, it specifically hydrolyzes proteins and peptide bonds C-terminally of Iysine and arginine. Amide and ester bonds of Arg and Lys are also cleaved. The specificity of Trypsin Sequencing Grade, modified, is verified with the oxidized B-chain of insulin (insulin Box) as a substrate. High concentrations of Trypsin Sequencing Grade, modified, one part by weight enzyme with 9 parts by weight insulin Box, are incubated for 18 hours to detect traces ofchymotrypsin impurities.

Preparation Note

Working concentration: 1/100 to 1/5 of the protein by weight
Storage conditions (working solution): -15 to -25 °C
Trypsin Sequencing Grade, modified, is more resistant to autolysis, even at pH values in the neutral and weakly basic range. The enzyme can be used in high concentrations.
A solution in 1% acetic acid or 1 mM HCI can be used for up to one week when stored at 2 to 8° C. Stored in aliquots at -15 to -25 °C, the solution is stable for at least one year without loss of activity.
Store dry

Analysis Note

Purity: Free of impurities that may interfere with the separation of peptides in reversed-phase HPLC.

Other Notes

For life science research only. Not for use in diagnostic procedures.


pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
1141803300104061838250834
1141802500104061838250827