Skip to Content
Merck
All Photos(1)

Key Documents

S1378

Sigma-Aldrich

Secobarbital sodium salt

Synonym(s):

5-Allyl-5-(1-methylbutyl)barbituric acid sodium-potassium salt, Quinalbarbitone sodium-potassium salt, Seconal sodium-potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N2NaO3
CAS Number:
Molecular Weight:
260.26
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: soluble
alcohol: soluble

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

SMILES string

[Na+].CCCC(C)C1(CC=C)C(=O)NC([O-])=NC1=O

InChI

1S/C12H18N2O3.Na/c1-4-6-8(3)12(7-5-2)9(15)13-11(17)14-10(12)16;/h5,8H,2,4,6-7H2,1,3H3,(H2,13,14,15,16,17);/q;+1/p-1

InChI key

AXXJTNXVUHVOJW-UHFFFAOYSA-M

Biochem/physiol Actions

Sedative; hypnotic.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B Kamien et al.
Pharmacology, biochemistry, and behavior, 58(4), 983-991 (1998-01-04)
Humans were trained to discriminate the benzodiazepine triazolam (0.32 mg/70 kg) from placebo under a two-response (drug vs. placebo) drug discrimination procedure. Dose-effect curves for several drugs were then determined in a crossover design using the two-response procedure and a
Mathilde Couteaudier et al.
Virology, 541, 25-31 (2019-12-13)
Simian foamy viruses (SFVs) are complex retroviruses that are widespread throughout nonhuman primates. SFVs can also be transmitted to humans, mostly through bites. We previously observed that primary zoonotic gorilla SFV strains grow much more slowly than laboratory-adapted chimpanzee strains.
Bruno Pinto et al.
Neuron, 108(5), 887-904 (2020-10-08)
Microglia are brain-resident immune cells and regulate mechanisms essential for cognitive functions. Down syndrome (DS), the most frequent cause of genetic intellectual disability, is caused by a supernumerary chromosome 21, containing also genes related to the immune system. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service