Skip to Content
Merck
All Photos(1)

Key Documents

S0202

Sigma-Aldrich

Secoisolariciresinol diglucoside

≥97% (HPLC)

Synonym(s):

SDG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H46O16
CAS Number:
Molecular Weight:
686.70
UNSPSC Code:
12352205
NACRES:
NA.25

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

protect from light

color

white to off-white

solubility

H2O: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O3[C@H]([C@@H]([C@H]([C@@H]([C@H]3CO)O)O)O)O.O(C)c1c(ccc(c1)C[C@H]([C@H](CO)Cc2cc(c(cc2)O)OC)CO)O

InChI

1S/C20H26O6.C6H12O6/c1-25-19-9-13(3-5-17(19)23)7-15(11-21)16(12-22)8-14-4-6-18(24)20(10-14)26-2;7-1-2-3(8)4(9)5(10)6(11)12-2/h3-6,9-10,15-16,21-24H,7-8,11-12H2,1-2H3;2-11H,1H2/t15-,16-;2-,3-,4+,5-,6-/m01/s1

InChI key

MJYQFWSXKFLTAY-OVEQLNGDSA-N

General description

Secoisolariciresinol diglucoside (SDG) is one of the essential dietary lignans found in flaxseed. It has an oligomeric structure.

Application


  • Supplementing flaxseed meal with sucrose, flaxseed oil, or both: Effect on milk enterolactone, ruminal microbiota profile, production performance, and nutrient utilization in dairy cows.: This study explores the effects of secoisolariciresinol diglucoside (SDG) from flaxseed meal on milk production, enterolactone levels, and the ruminal microbiota profile in dairy cows, highlighting its role in improving nutrient utilization and production performance (Ghedini CP et al., 2024).

  • Successful Outcome of a Patient with Concomitant Pancreatic and Renal Carcinoma Receiving Secoisolariciresinol Diglucoside Therapy Alone: A Case Report.: This case report documents a patient with pancreatic and renal carcinoma treated solely with SDG, showcasing its potential therapeutic effects in oncology and highlighting a novel application in cancer treatment (Wu H et al., 2024).

  • Linagliptin and secoisolariciresinol diglucoside attenuate hyperlipidemia and cardiac hypertrophy induced by a high-methionine diet in rats via suppression of hyperhomocysteinemia-induced endoplasmic reticulum stress.: This study demonstrates the combined therapeutic effects of SDG and linagliptin in reducing hyperlipidemia and cardiac hypertrophy, highlighting its potential in cardiovascular health applications (Jalal IA et al., 2023).

Biochem/physiol Actions

Secoisolariciresinol diglucoside (SDG) a metal-chelating ligand, that displays antiproliferative and proapoptotic effects in breast cancer cells. It elicits reno-protective properties against cadmium-induced nephrotoxicity in rats. SDG is a potent anti-inflammatory agent which prevents the interaction of blood-brain-barrier with inflammatory cells and protects against neuroinflammatory diseases. It is also an excellent anti-diabetic agent.
Secoisolariciresinol diglucoside is a phytochemical present in flax, sunflower, sesame and pumpkin seeds. It is a precursor of mammal lignans and a phytoestrogen. Secoisolariciresinol diglucoside has been shown to have antioxidant and cardioprotective properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Márcia Constantino Colli et al.
Journal of medicinal food, 15(9), 840-845 (2012-08-29)
The objective of this study was to evaluate the efficacy of flaxseed meal and flaxseed extract in reducing climacteric symptoms of menopausal women. Ninety menopausal women were randomly distributed into three study groups: group I received 1 g per day of
Parfait Kezimana et al.
Frontiers in genetics, 9, 641-641 (2019-01-09)
Secoisolariciresinol diglucoside (SDG), found mainly in flaxseed, is one of the essential lignans. SDG, as well as the beneficial fatty acid composition and high fiber content, has made flaxseed an important source of functional food or nutraceutical ingredients. Various studies
Thomas J Schmidt et al.
Phytochemistry, 82, 89-99 (2012-08-03)
Mature seeds of 20 Linum species were analyzed for their content of lignans. The seeds of common flax (Linum usitatissimum L.) are known to contain as characteristic lignan sesoisolariciresinol diglucoside (SDG), whose presence in seeds of some other Linum species
Xian-wei Li et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 35(2), 248-253 (2012-07-25)
To study the influence of Secoisolariciresinol Diglucoside (SDG) combined with Bortezomib on induction of apoptosis in lung cancer cell line A549 and its relative mechanisms. The effect on proliferation was evaluated by MTT assay. The cell apoptosis was studied by
Kailash Prasad
Current pharmaceutical biotechnology, 11(8), 794-800 (2010-09-30)
Many natural products, including vitamin E, garlic, purpurogallin, flaxseed and its components [secoisolariciresinol diglucoside (SDG) and flax lignan complex (FLC)] and resveratrol have been reported to suppress hypercholesterolemic atherosclerosis. It is known that all of the drugs that suppress the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service