RAB0256
Human IL-13 ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type urine
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.15 pg/mL
standard curve range: 0.16-40 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL13(3596)
General description
The Human IL-13 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-13 in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human IL-13
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Arvind Chopra et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(2), 319-326 (2014-02-08)
To evaluate whether chloroquine (CQ) is more effective than meloxicam for treating early musculoskeletal pain and arthritis following acute chikungunya (CHIK) virus infection. During the 2006 CHIK epidemic, 509 rural community cases of acute CHIK virus infection were identified in
Christian Schwartz et al.
Journal of immunology (Baltimore, Md. : 1950), 193(7), 3590-3599 (2014-08-31)
Schistosomiasis is a severe and chronic disease caused by the parasitic trematode Schistosoma mansoni after deposition of eggs in the liver and intestines. The immune response to S. mansoni eggs is characterized by increased Th2 cells, eosinophilia, and high serum
Tae Young Jang et al.
International immunopharmacology, 22(2), 444-450 (2014-08-12)
To evaluate the antiallergic effects of oral benzaldehyde in a murine model of allergic asthma and rhinitis, we divided 20 female BALB/c mice aged 8-10 weeks into nonallergic (intraperitoneally sensitized and intranasally challenged to normal saline), allergic (intraperitoneally sensitized and
Eun-Hyeong Shim et al.
The Journal of pharmacy and pharmacology, 66(9), 1303-1316 (2014-06-26)
We aimed to investigate the inhibitory efficacy of Solanum tuberosum L. var. Vitelotte (SV) extract on atopic dermatitis (AD)-like skin lesions induced by the topical application of 2,4-dinitrochlorobenzene in NC/Nga mice. SV extract was administered orally to NC/Nga mice at the
P N Adams et al.
Parasite immunology, 36(10), 531-539 (2014-07-22)
The M2 subset of macrophages has a critical role to play in host tissue repair, tissue fibrosis and modulation of adaptive immunity during helminth infection. Infection with the helminth, Fasciola hepatica, is associated with M2 macrophages in its mammalian host
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service