Skip to Content
Merck
All Photos(1)

Key Documents

RAB0506

Sigma-Aldrich

Mouse VCAM-1 ELISA Kit

for serum, plasma, cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 2.05-500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Vcam1(22329)

General description

The Mouse VCAM-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse VCAM-1 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse VCAM1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Zoladz et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 65(3), 441-448 (2014-06-17)
It has been demonstrated that physical training increases serum brain-derived neurotrophic factor (BDNF) in healthy people. The aim of this study was to establish the effect of physical training on the basal serum level of the BDNF in the Parkinson's
Martha Lappas
Metabolism: clinical and experimental, 63(6), 860-873 (2014-04-02)
To determine the effect of maternal obesity and gestational diabetes mellitus (GDM) on the expression and release of genes involved in endothelial cell dysfunction in human placenta and omental adipose tissue. Human placenta and omental adipose tissue were obtained from
P Mierzejewska et al.
Molecular and cellular biochemistry, 458(1-2), 133-142 (2019-05-17)
Changes in the ecto-5'-nucleotidase activity-an extracellular nucleotide catabolic enzyme may lead to the inflammation and endothelial dysfunction. We investigated the effect of CD73 deletion on the endothelial function and L-arginine metabolism in various age groups of mice. 1-,3-,6-, and 12-month-old
Han Saem Jeong et al.
Phytotherapy research : PTR, 28(10), 1492-1498 (2014-04-08)
Black raspberry (Rubus occidentalis) has been known for its anti-inflammatory and anti-oxidant effects. However, short-term effects of black raspberry on lipid profiles and vascular endothelial function have not been investigated in patients with metabolic syndrome. Patients with metabolic syndrome (n = 77)
Isabella Werner et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 63(8), 649-656 (2014-05-06)
As methylene blue (MB) has been recently proposed to preserve blood pressure in case of vasoplegic syndrome and shock, an entity directly related to systemic inflammation, we aimed to elucidate the effect of MB on the expression of adhesion-molecules in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service