RAB0126
Human BLC / BCA-1 / CXCL13 ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type urine
sample type plasma
sample type cell culture supernatant(s)
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 1.37-1000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CXCL13(10563)
General description
The CXCL13 (C-X-C motif chemokine ligand 13 ) gene codes for a CXC subtype of the chemokine superfamily and it is also known as B cell-attracting chemokine 1. The CXCL13 gene is mapped to human chromosome 4q21.1.
Immunogen
Recombinant Human BLC
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
CXCL13 (C-X-C motif chemokine ligand 13 ) contributes to the process of tumorigenesis in breast cancer, neuronal cancer and prostatic cancer. CXCL13 serves as an important marker for Lyme neuroborreliosis. CXCL13 mediates B-cell migration,survival, and maturation. CXCL13 is a ligand for CXCR5 and possesses immune function such as antibody production.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
MDC and BLC are independently associated with the significant risk of early stage lung adenocarcinoma.
Zhang Y, et al.
Oncotarget, 7(50), 83051-83051 (2016)
Biomarker dynamics in B-cell lymphoma: a longitudinal prospective study of plasma samples up to 25 years before diagnosis.
Spath F, et al.
Cancer Research (2017)
Chemokine/chemokine receptor nomenclature.
Bacon K, et al.
Journal of Leukocyte Biology, 70, 465-466 (2001)
Elevated C?X?C motif ligand 13 and B?cell?activating factor levels in neuromyelitis optica during remission.
Wang S, et al.
Brain and Behavior, 7(4), e00648-e00648 (2017)
Cerebrospinal fluid B-lymphocyte chemoattractant CXCL13 in the diagnosis of acute Lyme neuroborreliosis in children.
Barstad B, et al.
The Pediatric Infectious Disease Journal, 36(12), e286-e292 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service