RAB0026
Human BDNF ELISA Kit
for serum, plasma, cell culture supernatant and urine
Synonym(s):
BDNF, Brain-derived neurotrophic factor
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type plasma
sample type serum
sample type cell culture supernatant(s)
sample type urine
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/mL
standard curve range: 0.066-16 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... BDNF(627)
General description
The Human BDNF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human BDNF in serum, plasma, cell culture supernatants and urine.
The brain derived neurotrophic factor (BDNF) gene is mapped to human chromosome 11p14.1. BDNF is a member of the neurotrophin family of growth factors. The gene encodes a precursor protein, proBDNF. Mature BDNF (mBDNF) is synthesized by post-translational cleavage of proBDNF. Both proBDNF and mBDNF play crucial roles in cellular signaling.
Immunogen
Recombinant Human BDNF
Application
For research use only. Not for use in diagnostic procedures.
Human BDNF ELISA kit has been used to determine the effect of bone marrow mesenchymal stem cell (MSC) transplantation on BDNF protein expression in the striatum of Tourette syndrome (TS) rats. It has also been used to determine the plasma BDNF levels in GD1 (Gaucher′s disease type 1) patients and control population.
Human BDNF ELISA kit has been used to determine the effect of bone marrow mesenchymal stem cell (MSC) transplantation on BDNF protein expression in the striatum of Tourette syndrome (TS) rats. It has also been used to determine the plasma BDNF levels in GD1 (Gaucher′s disease type 1) patients and control population.
Human BDNF ELISA Kit has been used:
- to measure the brain-derived neurotrophic factor (BDNF) protein expression
- to determine BDNF level in the slice culture medium and the extracts of organotypic brain slice cultures (OBSCs)
- to determine BDNF level in plasma
Biochem/physiol Actions
Brain derived neurotrophic factor (BDNF) is involved in hippocampal function and verbal episodic memory in humans. Hence, variation in the BDNF gene expression alters these neurological functions. BDNF also plays a vital role in vascular function and participates in angiogenesis via the specific receptor tropomyosin-related kinase B (TrkB). It is involved in the pathogenesis of Alzheimer′s disease. ProBDNF interacts with p75 neurotrophin receptor, leading to long-term depression in the hippocampus.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Oxidative Stress-Tolerant Stem Cells from Human Exfoliated Deciduous Teeth Decrease Hydrogen Peroxide-Induced Damage in Organotypic Brain Slice Cultures from Adult Mice
Xiao L, et al.
International Journal of Molecular Sciences, 20(8), 1858-1858 (2019)
Xiumei Liu et al.
Experimental and therapeutic medicine, 11(4), 1211-1216 (2016-04-14)
The aim of the present study was to investigate the effect of bone marrow mesenchymal stem cell (MSC) transplantation on brain-derived neurotrophic factor (BDNF) expression in the striatum of Tourette syndrome (TS) rats. In addition, the possible mechanism of MSC
Shervin Pejhan et al.
Frontiers in cell and developmental biology, 8, 763-763 (2020-09-26)
Rett Syndrome (RTT) is a rare and progressive neurodevelopmental disorder that is caused by de novo mutations in the X-linked Methyl CpG binding protein 2 (MECP2) gene and is subjected to X-chromosome inactivation. RTT is commonly associated with neurological regression
Effect of mesenchymal stem cell transplantation on brain-derived neurotrophic factor expression in rats with Tourette syndrome
Liu X, et al.
Experimental and Therapeutic Medicine, 11(4), 1211-1216 (2016)
Hélio José Coelho-Júnior et al.
International journal of environmental research and public health, 17(10) (2020-05-20)
The present study compared the effects of traditional resistance training (TRT) and combined power training (PT) and TRT (PTRT) on cognitive parameters and serum brain-derived neurotrophic factor (BDNF) levels in non-demented, well-functioning, community-dwelling older women. Forty-five older women were randomized
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service