Skip to Content
Merck
All Photos(1)

Key Documents

R7644

Sigma-Aldrich

Transcript RNA Markers 0.28-6.6 kb

for RNA electrophoresis

Synonym(s):

0.28-6.6 kb Transcript RNA Markers, Transcript RNA markers for RNA electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105337
NACRES:
NA.25

grade

Molecular Biology
for molecular biology

Quality Level

form

liquid

usage

25 uses

suitability

suitable for electrophoresis (RNA)

storage temp.

−70°C

General description

Sigma′s Transcript RNA Marker (0.28 – 6.6 kb) contains 9 RNA transcripts from 281 – 6,583 bases. Three μl of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose gel.

Application

Transcript RNAMarkers 0.28-6.6 kb are suitable for use as an RNA ladder for formaldehyde agarose gel electrophoresis. It is also suitable for the size determination of small RNA using native or denaturing agarose gel electrophoresis.

Components

Sigma′s Transcript RNA Marker (0.28 – 6.6 kb) is provided in a gel-loading solution with 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.

Other Notes

Contains a mixture of nine RNA transcripts, 281–6,583 bases
For optimal resolution, the recommended agarose gel concentration is 1.0%.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of two mRNA species at single-cell resolution by double-fluorescence in situ hybridization.
Pinaud R, et al.
Nature Protocols, 3, 1370-1379 (2008)
Characterization of M116. 1p, a murine cytomegalovirus protein required for efficient infection of mononuclear phagocytes
Ruzic T, et al.
Journal of Virology, 96(2), e00876-e00821 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service