Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
usage
sufficient for 100 sample(s) (per 500 µL of reagent)
feature
Intended use (For stabilization of RNA samples for RNA gel electrophoresis)
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
DNA purification: suitable
test parameters
sample volume: 10 μL per lane
greener alternative category
storage temp.
2-8°C
General description
Preparation Note
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description 2X Loading dye for RNA gel electrophoresis. 500µL for 100 samples. | description Reagents and consumables for clean-up of pre-purified DNA. 10, 50 or 250 purifications. | description for NA electrophoresis | description Reagents and consumables for organic solvent clean-up from RNA. 10, 50 or 250 purifications. |
| technique(s) DNA purification: suitable | technique(s) DNA purification: suitable | technique(s) - | technique(s) RNA purification: suitable |
| storage temp. 2-8°C | storage temp. room temp | storage temp. room temp | storage temp. room temp |
| greener alternative category | greener alternative category | greener alternative category - | greener alternative category |
| greener alternative product characteristics Waste Prevention | greener alternative product characteristics Waste Prevention | greener alternative product characteristics - | greener alternative product characteristics Waste Prevention |
| sustainability Greener Alternative Product | sustainability Greener Alternative Product | sustainability - | sustainability Greener Alternative Product |
| test parameters sample volume: 10 μL per lane | test parameters sample volume: 90-110 μL, : 3 min hands on time | test parameters - | test parameters sample volume: 90-110 μL, : 3 min hands on time |
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
supp_hazards
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






