Skip to Content
Merck
All Photos(1)

Key Documents

PHR1188

Supelco

Isotretinoin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

13-cis-Retinoic acid, Isotretinoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. I0800000
traceable to USP 1353500

API family

isotretinoin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

172-175 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

[H]\C(C(O)=O)=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C20H28O2/c1-15(8-6-9-16(2)14-19(21)22)11-12-18-17(3)10-7-13-20(18,4)5/h6,8-9,11-12,14H,7,10,13H2,1-5H3,(H,21,22)/b9-6+,12-11+,15-8+,16-14-

InChI key

SHGAZHPCJJPHSC-XFYACQKRSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Isotretinoin is a vitamin A derivative, obtained from the isomerization of retinoic acid in the body. It is therapeutically used to treat severe recalcitrant nodular acne. It is also reported to exhibit anti-inflammatory and anti-tumor activities.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Isotretinoin may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using reversed-phase high-performance liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

13-cis-Retinoic acid (RA) has anti-inflammatory and anti-tumor action. The action of RA is mediated through RAR-β and RAR-α receptors. RA attenuates iNOS expression and activity in cytokine-stimulated murine mesangial cells. It induces mitochondrial membrane permeability transition, observed as swelling and as a decrease in membrane potential, and stimulates the release of cytochrome c implicating mechanisms through the apoptosis pathway. These activities are reversed by EGTA and cyclosporin A. RA also increases MMP-1 protein expression partially via increased transcription.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0144 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of isotretinoin in pharmaceutical formulations by reversed-phase HPLC
Guimaraes AC, et al.
Journal of Biomedical Science and Engineering, 3, 454-458 (2010)
Kaisar Raza et al.
Colloids and surfaces. B, Biointerfaces, 105, 67-74 (2013-01-30)
Isotretinoin (ITR) is a drug of choice in the treatment of all types of acne, including recalcitrant, severe and nodulocystic. The most widely employed route of its administration, i.e., oral intake, is reported to be associated with severe side-effects including
F L Meyskens et al.
Critical reviews in oncology/hematology, 3(1), 75-101 (1985-01-01)
Retinoids, particularly 13-cis-retinoic acid, have shown great promise against a number of benign, but serious dermatological conditions. In animal models, 13-cis-retinoic acid functions is a potent antipromoter whether a cancer has been initiated by chemical, physical, or viral agents. Additionally
H J M J Crijns et al.
The British journal of dermatology, 164(2), 238-244 (2010-08-19)
Most of the publications on isotretinoin, pregnancy and compliance with the pregnancy prevention programme (PPP) originate from North America. Information specific for the European situation is very limited. The aim of this study was to identify publications describing the use
Navin Pinto et al.
Pediatric blood & cancer, 61(6), 1104-1106 (2013-12-03)
Patients <12 months with favorable biology, metastatic neuroblastoma have >90% overall survival following treatment with chemotherapy and surgery. We report two infants with favorable biology, stage 4 neuroblastoma with refractory disease after standard intermediate-risk chemotherapy and additional retrieval chemotherapy. One

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service