R1149
Resuspension Solution
Add RNase A before use. After addition of RNAse, store at 2-8ºC.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.25
Recommended Products
grade
Molecular Biology
for molecular biology
form
liquid
description
Add RNase A before use. After addition of RNAse, store at 2-8ºC.
storage temp.
room temp
General description
This resuspension buffer is for use with GenElute™ kits during sample preparation. The GenElute™ Plasmid Miniprep Kit offers a simple, rapid, and cost-effective method for isolating plasmid DNA from recombinant E. coli cultures
Legal Information
GenElute is a trademark of Sigma-Aldrich Co. LLC
sample prep
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Keiji Ueno et al.
Chemistry Central journal, 5(1), 16-16 (2011-04-06)
We have previously reported on the variation of total fructooligosaccharides (FOS), total inulooligosaccharides (IOS) and inulin in the roots of burdock stored at different temperatures. During storage at 0°C, an increase of FOS as a result of the hydrolysis of
Masa-aki Yoshida et al.
BMC evolutionary biology, 11, 180-180 (2011-06-28)
Coleoid cephalopods (squids and octopuses) have evolved a camera eye, the structure of which is very similar to that found in vertebrates and which is considered a classic example of convergent evolution. Other molluscs, however, possess mirror, pin-hole, or compound
Isabella Monne et al.
Journal of clinical microbiology, 46(5), 1769-1773 (2008-03-28)
Among the different hemagglutinin (HA) subtypes of avian influenza (AI) viruses, H5, H7, and H9 are of major interest because of the serious consequences for the poultry industry and the increasing frequency of direct transmission of these viruses to humans.
Merijn L M Salverda et al.
PLoS genetics, 7(3), e1001321-e1001321 (2011-03-17)
Whether evolution is erratic due to random historical details, or is repeatedly directed along similar paths by certain constraints, remains unclear. Epistasis (i.e. non-additive interaction between mutations that affect fitness) is a mechanism that can contribute to both scenarios. Epistasis
Aarti Kathrani et al.
PloS one, 7(1), e30117-e30117 (2012-01-27)
Single nucleotide polymorphisms (SNP) in the TLR5 gene have been associated with human inflammatory bowel disease (IBD) and animal models of this disease. We recently demonstrated a significant association between three non-synonymous SNPs in the canine TLR5 gene and IBD
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service