Skip to Content
Merck
All Photos(8)

Key Documents

NA0300

Sigma-Aldrich

GenElute HP Plasmid Maxiprep Kit

greener alternative

sufficient for 10 purifications

Synonym(s):

GenElute HP Plasmid Maxiprep Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

sufficient for 10 purifications

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

DNA purification: suitable

greener alternative category

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute HP Plasmid Maxiprep Kit offers a simple, rapid, and cost effective method for isolating plasmid DNA from recombinant E. coli cultures. The kit features a filter syringe for the rapid clearing of lysate and a silica binding column designed for either a vacuum or a spin format. Up to 1.2 mg of plasmid DNA can be isolated from a 150 mL overnight culture grown in Luria Broth (LB) medium. Note that the actual yield depends on the strain, the plasmid, and the culture medium used.

An overnight recombinant E. coli culture is harvested by centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the DNA onto a silica membrane in the presence of high salts. Contaminants are removed by two wash steps. Finally, the bound DNA is eluted in Elution Solution (Tris-HCl) or water.

The recovered plasmid DNA is predominately in its supercoiled form. The DNA is ready for immediate use in downstream applications such as restriction digestion, ligation, sequencing, PCR, transformation, and transfection.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Application

GenElute HP Plasmid Maxiprep Kit has been used in plasmid DNA extraction of SF91 or MIEV vector, apolipoprotein A-V mutant constructs, lentiviral vector pLenti6/R4R2/V5-DEST,metastasis-associated protein 1 (MTA1) and adipokinetic hormone receptor

Features and Benefits

  • From harvested bacterial culture to pure plasmid DNA in 30 minutes or less
  • Up to 1.2 mg of high-copy plasmid DNA
  • Flexibility of a vacuum or spin format
  • Contains fewer plastic components than other high speed kits, reducing the amount of waste
  • No phenol/chloroform extraction or alcohol precipitation required

Other Notes

For additional information, please see www.sigma-aldrich.com/genelutehp.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-oral cancer effects of triptolide by downregulation of DcR3 in vitro, in vivo, and in preclinical patient-derived tumor xenograft model
Yang CY, et al.
Head & Neck (2018)
Cellular barcoding tool for clonal analysis in the hematopoietic system
Gerrits A, et al.
Blood, blood-b2009 (2010)
Analysis of Peptide Ligand Specificity of Different Insect Adipokinetic Hormone Receptors
Marchal E, et al.
International Journal of Molecular Sciences, 19(2), 542-542 (2018)
A stem cell-like chromatin pattern may predispose tumor suppressor genes to DNA hypermethylation and heritable silencing
Ohm JE, et al.
Nature Genetics, 39(2), 237-237 (2007)
Determination of the functionality of common APOA5 polmorphisms
Talmud PJ, et al.
The Journal of Biological Chemistry (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service